Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ASTN2
Transcript:
NM_012210.3
Assay Name:
Mutation genome location:
chr.9 116697897-116698493 on build GRCh38
Assay transcription exon location:
Within Exon 2
Gene:
TRIM32
Transcript:
NM_012210.3
Assay Name:
Mutation genome location:
chr.9 116697897-116698493 on build GRCh38
Assay transcription exon location:
Within Exon 2
Assay ID
Hs00440588_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein localization to cell surface
protein polyubiquitination
tissue homeostasis
actin ubiquitination
response to UV
protein ubiquitination
positive regulation of cell growth
positive regulation of cell migration
regulation of type I interferon production
negative regulation of viral transcription
response to tumor necrosis factor
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
fat cell differentiation
positive regulation of neuron differentiation
positive regulation of protein catabolic process
positive regulation of cell cycle
positive regulation of proteolysis
muscle cell cellular homeostasis
negative regulation of fibroblast proliferation
positive regulation of neurogenesis
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
positive regulation of striated muscle cell differentiation
axon development
negative regulation of viral release from host cell
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
positive regulation of tumor necrosis factor-mediated signaling pathway
positive regulation of interleukin-17-mediated signaling pathway
positive regulation of chemokine (C-C motif) ligand 20 production
positive regulation of cell motility
calcium ion binding
inositol 1,3,4,5 tetrakisphosphate binding
transcription coactivator activity
RNA binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
myosin binding
Tat protein binding
translation initiation factor binding
ubiquitin binding
protein self-association
ubiquitin protein ligase activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline