Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
BTK | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_000061.2 | NP_000052.1 | - | - | - |
NM_001287344.1 | NP_001274273.1 | - | - | - | |
NM_001287345.1 | NP_001274274.1 | - | - | - | |
GenBank | AF153364.1 | AAF61477.1 | - | - | - |
AF153756.1 | AAF37333.1 | - | - | - | |
AF153757.1 | AAF37334.1 | - | - | - | |
AF153758.1 | AAF37335.1 | - | - | - | |
AF153759.1 | AAF37336.1 | - | - | - | |
AF153760.1 | AAF37337.1 | - | - | - | |
AF153761.1 | AAF37338.1 | - | - | - | |
AF153762.1 | AAF37339.1 | - | - | - | |
AF153764.1 | AAF37341.1 | - | - | - | |
AJ888376.1 | CAI59842.1 | - | - | - | |
AJ888377.1 | CAI59836.1 | - | - | - | |
AJ888378.1 | CAI59837.1 | - | - | - | |
AJ888379.1 | CAI59838.1 | - | - | - | |
AJ888380.1 | CAI59839.1 | - | - | - | |
AJ888381.1 | CAI59840.1 | - | - | - | |
AK057105.1 | - | - | - | ||
AK225577.1 | - | - | - | ||
AK309851.1 | - | - | - | ||
AK314382.1 | - | - | - | ||
AM051275.1 | CAJ19681.1 | - | - | - | |
AM051276.1 | CAJ19682.1 | - | - | - | |
AM051277.1 | CAJ19683.1 | - | - | - | |
AM051278.1 | CAJ19684.1 | - | - | - | |
AM051279.1 | CAJ19685.1 | - | - | - | |
AM051280.1 | CAJ19686.1 | - | - | - | |
AM051281.1 | CAJ19687.1 | - | - | - | |
AM051282.1 | CAJ19688.1 | - | - | - | |
AM051283.1 | CAJ19689.1 | - | - | - | |
AM051284.1 | CAJ19690.1 | - | - | - | |
AM051285.1 | CAJ19691.1 | - | - | - | |
AM051286.1 | CAJ19692.1 | - | - | - | |
BC109079.1 | AAI09080.1 | - | - | - | |
BC109080.1 | AAI09081.1 | - | - | - | |
ENST00000308731.0 | - | - | - | ||
KF241986.1 | - | - | - | ||
X58957.1 | CAA41728.1 | - | - | - |
Literature Links: |
BTK PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Less than 200 bp |