Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
HSP90AA1
Transcript:
NM_001017963.2
Assay Name:
Mutation genome location:
chr.14 102081664-102081830 on build GRCh38
Assay ID
Hs00519466_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
Hsp90 family chaperone
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
neuron migration
skeletal muscle contraction
mitochondrial transport
receptor-mediated endocytosis
response to unfolded protein
signal transduction
response to heat
response to cold
response to salt stress
positive regulation of lamellipodium assembly
cardiac muscle cell apoptotic process
peptidyl-tyrosine phosphorylation
regulation of protein ubiquitination
positive regulation of protein import into nucleus, translocation
Fc-gamma receptor signaling pathway involved in phagocytosis
ERBB2 signaling pathway
protein refolding
regulation of protein complex assembly
protein unfolding
response to estrogen
protein import into mitochondrial outer membrane
positive regulation of nitric oxide biosynthetic process
positive regulation of cell size
response to antibiotic
vascular endothelial growth factor receptor signaling pathway
protein stabilization
regulation of nitric-oxide synthase activity
chaperone-mediated protein complex assembly
positive regulation of cardiac muscle contraction
chaperone-mediated autophagy
regulation of cellular response to heat
nucleotide binding
UTP binding
CTP binding
mRNA binding
protein tyrosine kinase activity
protein binding
ATP binding
GTP binding
ATPase activity
sulfonylurea receptor binding
protein kinase binding
protein phosphatase binding
MHC class II protein complex binding
nitric-oxide synthase regulator activity
TPR domain binding
dATP binding
identical protein binding
protein homodimerization activity
histone deacetylase binding
ion channel binding
poly(A) RNA binding
GTPase binding
Rho GDP-dissociation inhibitor binding
unfolded protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
git-commit: c9e08c96761173abe34e68f880379696776a4827
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.1-2026.02.97.1.0