Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
CSMD3 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_052900.2 | NP_443132.3 | - | - | - |
NM_198123.1 | NP_937756.1 | - | - | - | |
NM_198124.1 | NP_937757.1 | 1137795 | c.10661G>T | p.G3554V | |
4979534 | c.10683T>C | p.Y3561Y | |||
3163174 | c.10687C>A | p.Q3563K | |||
1154900 | c.10678G>T | p.D3560Y | |||
1249256 | c.10647T>G | p.T3549T | |||
6455502 | c.10678G>A | p.D3560N | |||
6516516 | c.10708G>T | p.G3570* | |||
1150863 | c.10708+1G>T | p.? | |||
1700477 | c.10672G>A | p.G3558S | |||
4568308 | c.10680T>A | p.D3560E | |||
5455625 | c.10708+7T>C | p.? | |||
5583410 | c.10685G>A | p.G3562E | |||
6492535 | c.10634C>A | p.P3545H | |||
5873746 | c.10660G>A | p.G3554R | |||
4604037 | c.10679A>T | p.D3560V | |||
6179426 | c.10669C>A | p.Q3557K | |||
4150456 | c.10660G>C | p.G3554R | |||
4520901 | c.10707G>A | p.L3569L | |||
7363055 | c.10680T>G | p.D3560E | |||
6516518 | c.10701A>T | p.Q3567H | |||
5997843 | c.10634C>T | p.P3545L | |||
6472268 | c.10689A>G | p.Q3563Q | |||
1597418 | c.10665T>G | p.F3555L | |||
1178863 | c.10644T>A | p.A3548A | |||
5759142 | c.10679A>G | p.D3560G | |||
1249303 | c.10677A>C | p.K3559N | |||
4675073 | c.10702delA | p.R3568fs*4 | |||
XM_011516815.2 | XP_011515117.1 | - | - | - | |
XM_011516816.2 | XP_011515118.1 | - | - | - | |
XM_017013008.1 | XP_016868497.1 | - | - | - | |
XM_017013009.1 | XP_016868498.1 | 201485 | c.10798G>T | p.D3600Y | |
1249302 | c.10797A>C | p.K3599N | |||
1178862 | c.10764T>A | p.A3588A | |||
4979533 | c.10803T>C | p.Y3601Y | |||
4604036 | c.10799A>T | p.D3600V | |||
5583409 | c.10805G>A | p.G3602E | |||
360466 | c.10793G>T | p.G3598V | |||
5455624 | c.10827+8T>C | p.? | |||
4520900 | c.10827G>A | p.L3609L | |||
4675072 | c.10822delA | p.R3608fs*4 | |||
6455501 | c.10798G>A | p.D3600N | |||
6516515 | c.10827+1G>T | p.? | |||
1249255 | c.10767T>G | p.T3589T | |||
319713 | c.10807C>A | p.Q3603K | |||
5997842 | c.10754C>T | p.P3585L | |||
6179425 | c.10789C>A | p.Q3597K | |||
319707 | c.10781G>T | p.G3594V | |||
6492534 | c.10754C>A | p.P3585H | |||
1095039 | c.10785T>G | p.F3595L | |||
1700476 | c.10792G>A | p.G3598S | |||
7363054 | c.10800T>G | p.D3600E | |||
4150455 | c.10780G>C | p.G3594R | |||
6472267 | c.10809A>G | p.Q3603Q | |||
5873745 | c.10780G>A | p.G3594R | |||
748721 | c.10827+2G>T | p.? | |||
6516517 | c.10821A>T | p.Q3607H | |||
5759141 | c.10799A>G | p.D3600G | |||
4568307 | c.10800T>A | p.D3600E | |||
XM_017013010.1 | XP_016868499.1 | - | - | - | |
XM_017013011.1 | XP_016868500.1 | - | - | - | |
XM_017013012.1 | XP_016868501.1 | - | - | - | |
GenBank | AB067481.2 | BAB67787.2 | - | - | - |
AB114604.1 | BAC82443.1 | - | - | - | |
AB114605.1 | BAC82444.1 | - | - | - | |
AK095111.1 | - | - | - | ||
AY210419.1 | AAO34702.1 | - | - | - | |
ENST00000297405.0 | - | - | - |
Literature Links: |
CSMD3 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |