Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
INPP5K
Transcript:
NM_016532.3
Assay Name:
Mutation genome location:
chr.17 1497895-1498166 on build GRCh38
Assay ID
Hs00547580_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
negative regulation of protein phosphorylation
regulation of glycogen biosynthetic process
negative regulation of protein kinase activity
phosphatidylinositol biosynthetic process
G-protein coupled receptor signaling pathway
negative regulation of peptidyl-threonine phosphorylation
negative regulation of glucose transport
dephosphorylation
actin cytoskeleton organization
cellular response to insulin stimulus
cellular response to hormone stimulus
negative regulation of peptidyl-serine phosphorylation
negative regulation of dephosphorylation
positive regulation of urine volume
glucose homeostasis
negative regulation of MAP kinase activity
negative regulation by host of viral transcription
negative regulation of glycogen biosynthetic process
negative regulation of single stranded viral RNA replication via double stranded DNA intermediate
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of insulin receptor signaling pathway
inositol phosphate dephosphorylation
phosphatidylinositol dephosphorylation
negative regulation of stress fiber assembly
negative regulation of protein kinase B signaling
negative regulation of calcium ion transport
cellular response to cAMP
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
protein targeting to plasma membrane
negative regulation of protein targeting to membrane
ruffle assembly
negative regulation of glycogen (starch) synthase activity
positive regulation of renal water transport
phosphatidylinositol-4,5-bisphosphate 5-phosphatase activity
inositol-polyphosphate 5-phosphatase activity
vasopressin receptor activity
protein binding
inositol bisphosphate phosphatase activity
phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity
phosphatidylinositol trisphosphate phosphatase activity
phosphatidylinositol phosphate 5-phosphatase activity
lipid phosphatase activity
inositol trisphosphate phosphatase activity
inositol-1,4,5-trisphosphate 5-phosphatase activity
inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline