Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TLR9
Transcript:
NM_017442.3
Assay Name:
Mutation genome location:
chr.3 52221767-52222010 on build GRCh38
Assay transcription exon location:
Within Exon 2
Assay ID
Hs00554912_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Function(s)
Process(es)
toll-like receptor signaling pathway
response to molecule of bacterial origin
MyD88-dependent toll-like receptor signaling pathway
inflammatory response
I-kappaB phosphorylation
axonogenesis
positive regulation of gene expression
maintenance of gastrointestinal epithelium
negative regulation of NF-kappaB transcription factor activity
tumor necrosis factor production
negative regulation of interleukin-6 production
negative regulation of interleukin-8 production
positive regulation of chemokine production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-beta production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-18 production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
negative regulation of toll-like receptor signaling pathway
positive regulation of toll-like receptor signaling pathway
toll-like receptor 9 signaling pathway
phosphatidylinositol-3-phosphate biosynthetic process
positive regulation of NF-kappaB import into nucleus
defense response to bacterium
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JUN kinase activity
positive regulation of interferon-gamma biosynthetic process
innate immune response
positive regulation of interferon-alpha biosynthetic process
positive regulation of interferon-beta biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
regulation of cytokine secretion
positive regulation of inflammatory response
defense response to Gram-negative bacterium
positive regulation of NF-kappaB transcription factor activity
positive regulation of nitric-oxide synthase biosynthetic process
negative regulation of calcium-transporting ATPase activity
transmembrane signaling receptor activity
interleukin-1 receptor binding
1-phosphatidylinositol-3-kinase activity
siRNA binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0