Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
KRT8
Transcript:
NM_002273.3
Assay Name:
Mutation genome location:
chr.12 52901084-52901349 on build GRCh38
Assay ID
Hs00567982_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Function(s)
Process(es)
viral process
tumor necrosis factor-mediated signaling pathway
sarcomere organization
response to hydrostatic pressure
response to other organism
cell differentiation involved in embryonic placenta development
extrinsic apoptotic signaling pathway
hepatocyte apoptotic process
structural molecule activity
protein binding
protein complex binding
scaffold protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline