Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TRPV4
Transcript:
NM_021625.4
Assay Name:
Mutation genome location:
chr.12 109783552-109783809 on build GRCh38
Assay ID
Hs00573694_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
diet induced thermogenesis
calcium ion transport
cellular calcium ion homeostasis
cell volume homeostasis
actin filament organization
cell-cell junction assembly
positive regulation of cytosolic calcium ion concentration
osmosensory signaling pathway
response to mechanical stimulus
positive regulation of gene expression
positive regulation of macrophage chemotaxis
negative regulation of neuron projection development
vasopressin secretion
positive regulation of microtubule depolymerization
actin cytoskeleton reorganization
response to insulin
cellular response to heat
hyperosmotic salinity response
glucose homeostasis
positive regulation of vascular permeability
cortical microtubule organization
positive regulation of JNK cascade
microtubule polymerization
regulation of response to osmotic stress
positive regulation of inflammatory response
multicellular organismal water homeostasis
positive regulation of ERK1 and ERK2 cascade
calcium ion import
calcium ion transmembrane transport
cellular response to osmotic stress
cellular hypotonic response
cellular hypotonic salinity response
positive regulation of monocyte chemotactic protein-1 production
positive regulation of macrophage inflammatory protein 1 alpha production
positive regulation of chemokine (C-C motif) ligand 5 production
blood vessel endothelial cell delamination
negative regulation of brown fat cell differentiation
signal transduction involved in regulation of aerobic respiration
positive regulation of chemokine (C-X-C motif) ligand 1 production
positive regulation of energy homeostasis
positive regulation of interleukin-6 secretion
actin binding
osmosensor activity
protein kinase C binding
cation channel activity
calcium channel activity
protein binding
calmodulin binding
ATP binding
microtubule binding
stretch-activated, cation-selective, calcium channel activity
protein kinase binding
SH2 domain binding
alpha-tubulin binding
beta-tubulin binding
actin filament binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline