Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CACNA1A
Transcript:
NM_001127222.1
Assay Name:
Mutation genome location:
chr.19 13210492-13210763 on build GRCh38
Assay ID
Hs00696472_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
sulfur amino acid metabolic process
glucose metabolic process
positive regulation of cytosolic calcium ion concentration
gamma-aminobutyric acid signaling pathway
neuromuscular synaptic transmission
synapse assembly
adult walking behavior
cell death
gamma-aminobutyric acid secretion
regulation of acetylcholine secretion, neurotransmission
cell growth
regulation of calcium ion-dependent exocytosis
transmission of nerve impulse
spinal cord motor neuron differentiation
cerebellum maturation
cerebellar molecular layer development
cerebellar Purkinje cell differentiation
vestibular nucleus development
cellular chloride ion homeostasis
negative regulation of hormone biosynthetic process
regulation of ion transmembrane transport
synaptic transmission, glutamatergic
neurotransmitter metabolic process
hormone metabolic process
receptor clustering
negative regulation of neuron apoptotic process
behavioral response to pain
calcium ion-regulated exocytosis of neurotransmitter
dendrite morphogenesis
regulation of axonogenesis
regulation of insulin secretion
musculoskeletal movement, spinal reflex action
neuromuscular process controlling balance
membrane depolarization
rhythmic synaptic transmission
calcium ion transmembrane transport
membrane depolarization during action potential
voltage-gated calcium channel activity
calcium channel activity
protein binding
high voltage-gated calcium channel activity
syntaxin binding
metal ion binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline