Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ANK3 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001149.3 | NP_001140.2 | 5814552 | c.1908A>G | p.S636S |
6586218 | c.1910T>A | p.V637E | |||
7337341 | c.1892G>A | p.R631K | |||
919221 | c.1889C>T | p.A630V | |||
NM_001204403.1 | NP_001191332.1 | - | - | - | |
NM_001204404.1 | NP_001191333.1 | - | - | - | |
NM_001320874.1 | NP_001307803.1 | 919220 | c.12326C>T | p.A4109V | |
6586217 | c.12347T>A | p.V4116E | |||
5814551 | c.12345A>G | p.S4115S | |||
7337340 | c.12329G>A | p.R4110K | |||
NM_020987.4 | NP_066267.2 | - | - | - | |
XM_005269715.3 | XP_005269772.1 | - | - | - | |
XM_006717796.3 | XP_006717859.1 | - | - | - | |
XM_006717802.3 | XP_006717865.1 | - | - | - | |
XM_011539708.2 | XP_011538010.1 | - | - | - | |
XM_011539709.2 | XP_011538011.1 | - | - | - | |
XM_017016110.1 | XP_016871599.1 | - | - | - | |
XM_017016111.1 | XP_016871600.1 | - | - | - | |
XM_017016112.1 | XP_016871601.1 | - | - | - | |
XM_017016113.1 | XP_016871602.1 | - | - | - | |
XM_017016114.1 | XP_016871603.1 | - | - | - | |
XM_017016115.1 | XP_016871604.1 | - | - | - | |
XM_017016116.1 | XP_016871605.1 | - | - | - | |
XM_017016117.1 | XP_016871606.1 | - | - | - | |
XM_017016118.1 | XP_016871607.1 | - | - | - | |
XM_017016119.1 | XP_016871608.1 | - | - | - | |
XM_017016120.1 | XP_016871609.1 | - | - | - | |
XM_017016121.1 | XP_016871610.1 | - | - | - | |
XM_017016122.1 | XP_016871611.1 | - | - | - | |
XM_017016123.1 | XP_016871612.1 | - | - | - | |
XM_017016124.1 | XP_016871613.1 | - | - | - | |
XM_017016125.1 | XP_016871614.1 | - | - | - | |
XM_017016126.1 | XP_016871615.1 | - | - | - | |
XM_017016127.1 | XP_016871616.1 | - | - | - | |
XM_017016128.1 | XP_016871617.1 | - | - | - | |
XM_017016129.1 | XP_016871618.1 | - | - | - | |
XM_017016130.1 | XP_016871619.1 | - | - | - | |
XM_017016131.1 | XP_016871620.1 | - | - | - | |
XM_017016132.1 | XP_016871621.1 | - | - | - | |
XM_017016133.1 | XP_016871622.1 | - | - | - | |
XM_017016134.1 | XP_016871623.1 | - | - | - | |
XM_017016135.1 | XP_016871624.1 | - | - | - | |
XM_017016136.1 | XP_016871625.1 | - | - | - | |
XM_017016137.1 | XP_016871626.1 | - | - | - | |
XM_017016138.1 | XP_016871627.1 | - | - | - | |
XM_017016139.1 | XP_016871628.1 | - | - | - | |
XM_017016140.1 | XP_016871629.1 | - | - | - | |
XM_017016141.1 | XP_016871630.1 | - | - | - | |
XM_017016142.1 | XP_016871631.1 | - | - | - | |
XM_017016143.1 | XP_016871632.1 | - | - | - | |
XM_017016144.1 | XP_016871633.1 | - | - | - | |
XM_017016145.1 | XP_016871634.1 | - | - | - | |
XM_017016146.1 | XP_016871635.1 | - | - | - | |
XM_017016147.1 | XP_016871636.1 | - | - | - | |
XM_017016148.1 | XP_016871637.1 | - | - | - | |
XM_017016149.1 | XP_016871638.1 | - | - | - | |
XM_017016150.1 | XP_016871639.1 | - | - | - | |
GenBank | AK092361.1 | - | - | - | |
AK126851.1 | BAC86721.1 | - | - | - | |
AK292927.1 | - | - | - | ||
AK295661.1 | - | - | - | ||
AL136710.1 | CAB66645.1 | - | - | - | |
BX537917.1 | CAD97900.2 | - | - | - | |
BX648574.1 | CAI56716.1 | - | - | - | |
ENST00000280772.0 | - | - | - | ||
U13616.1 | AAA64834.1 | - | - | - |
Literature Links: |
ANK3 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |