Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
HSP90AA1
Transcript:
NM_001017963.2
Assay Name:
Mutation genome location:
chr.14 102081986-102082247 on build GRCh38
Assay ID
Hs00725555_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
Hsp90 family chaperone
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
neuron migration
skeletal muscle contraction
mitochondrial transport
receptor-mediated endocytosis
response to unfolded protein
signal transduction
response to heat
response to cold
response to salt stress
positive regulation of lamellipodium assembly
cardiac muscle cell apoptotic process
peptidyl-tyrosine phosphorylation
regulation of protein ubiquitination
positive regulation of protein import into nucleus, translocation
Fc-gamma receptor signaling pathway involved in phagocytosis
ERBB2 signaling pathway
protein refolding
regulation of protein complex assembly
protein unfolding
response to estrogen
protein import into mitochondrial outer membrane
positive regulation of nitric oxide biosynthetic process
positive regulation of cell size
response to antibiotic
vascular endothelial growth factor receptor signaling pathway
protein stabilization
regulation of nitric-oxide synthase activity
chaperone-mediated protein complex assembly
positive regulation of cardiac muscle contraction
chaperone-mediated autophagy
regulation of cellular response to heat
nucleotide binding
UTP binding
CTP binding
mRNA binding
protein tyrosine kinase activity
protein binding
ATP binding
GTP binding
ATPase activity
sulfonylurea receptor binding
protein kinase binding
protein phosphatase binding
MHC class II protein complex binding
nitric-oxide synthase regulator activity
TPR domain binding
dATP binding
identical protein binding
protein homodimerization activity
histone deacetylase binding
ion channel binding
poly(A) RNA binding
GTPase binding
Rho GDP-dissociation inhibitor binding
unfolded protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
git-commit: c9e08c96761173abe34e68f880379696776a4827
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.1-2026.02.97.1.0