Hamburger Menu Button
Thermo Fisher Scientific Logo
로그인
회원이 아니신가요?
계정 생성하기
모든 제품 주문
항체
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR 분석
피펫 및 피펫 팁
실험실용 원심분리기
초저온 냉동고
분광학
비이커
PCR 장비 및 용품
제품 카테고리 전체보기
응용 분야 및 기법
세포 분석
실험실 장비
Real-Time PCR
PCR
크로마토그래피
세포 배양 및 트랜스펙션
DNA/RNA 추출과 분석
단백질 생물학
유세포분석
화학 제품
응용 분야 및 기법 전체보기
서비스
맞춤형 서비스
엔터프라이즈급 실험실 정보 처리
엔터프라이즈 서비스
360° CDMO 및 CRO 서비스
CDMO 서비스
임상 시험 CRO 서비
장비 서비스
교육 서비스
서비스 전체 보기
지원
고객센터
문의하기
시험성적서(COA) 및 적합성 인증서(COC)
Safety Data Sheets (SDS)
매뉴얼
인용 및 참고 문헌
기기 지원
기술 자료/제품 FAQ
학습 센터
지원 메뉴 전체보기
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
문의하기
빠른 주문
주문 현황
제품 문서 검색
Thermo Fisher Scientific Logo
Search
전체검색
전체검색
Search button
주문 현황
빠른주문
로그인
로그인
회원이 아니신가요?
계정 생성하기
계정정보
주문내역조회
커스텀제품 및 프로젝트
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TLR9
Transcript:
NM_017442.3
Assay Name:
Mutation genome location:
chr.3 52223439-52223702 on build GRCh38
Assay transcription exon location:
Within Exon 2
Assay ID
Hs00775435_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
재고 정보
주문접수 후 제작
Catalog #
A15629
제품 정가
Your Price
온라인 행사가:
공급가 확인 ›
카트에 추가
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay 상세 정보
Back To Top
추가 정보
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Function(s)
Process(es)
toll-like receptor signaling pathway
response to molecule of bacterial origin
MyD88-dependent toll-like receptor signaling pathway
inflammatory response
I-kappaB phosphorylation
axonogenesis
positive regulation of gene expression
maintenance of gastrointestinal epithelium
negative regulation of NF-kappaB transcription factor activity
tumor necrosis factor production
negative regulation of interleukin-6 production
negative regulation of interleukin-8 production
positive regulation of chemokine production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-beta production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-18 production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
negative regulation of toll-like receptor signaling pathway
positive regulation of toll-like receptor signaling pathway
toll-like receptor 9 signaling pathway
phosphatidylinositol-3-phosphate biosynthetic process
positive regulation of NF-kappaB import into nucleus
defense response to bacterium
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JUN kinase activity
positive regulation of interferon-gamma biosynthetic process
innate immune response
positive regulation of interferon-alpha biosynthetic process
positive regulation of interferon-beta biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
regulation of cytokine secretion
positive regulation of inflammatory response
defense response to Gram-negative bacterium
positive regulation of NF-kappaB transcription factor activity
positive regulation of nitric-oxide synthase biosynthetic process
negative regulation of calcium-transporting ATPase activity
transmembrane signaling receptor activity
interleukin-1 receptor binding
1-phosphatidylinositol-3-kinase activity
siRNA binding
Back To Top
관련 제품
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-xjq9k:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0