Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
SOX9
Transcript:
NM_000346.3
Assay Name:
Mutation genome location:
chr.17 72121345-72121600 on build GRCh38
Assay transcription exon location:
Within Exon 1
Assay ID
Hs00821908_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
skeletal system development
cartilage condensation
ossification
branching involved in ureteric bud morphogenesis
cell fate specification
epithelial to mesenchymal transition
tissue homeostasis
positive regulation of protein phosphorylation
hair follicle development
positive regulation of mesenchymal cell proliferation
negative regulation of immune system process
heart valve development
heart valve morphogenesis
heart valve formation
endocardial cushion morphogenesis
chondrocyte differentiation involved in endochondral bone morphogenesis
chondrocyte hypertrophy
nucleosome assembly
chromatin remodeling
transcription from RNA polymerase II promoter
protein complex assembly
cytoskeleton organization
signal transduction
epidermal growth factor receptor signaling pathway
Notch signaling pathway
spermatogenesis
positive regulation of cell proliferation
male gonad development
regulation of cell cycle process
positive regulation of epithelial cell migration
neural crest cell development
positive regulation of phosphatidylinositol 3-kinase signaling
single organismal cell-cell adhesion
male germ-line sex determination
cAMP-mediated signaling
regulation of cell adhesion
extracellular matrix organization
negative regulation of ossification
negative regulation of bone mineralization
prostate gland development
negative regulation of epithelial cell differentiation
positive regulation of epithelial cell differentiation
mammary gland development
notochord development
otic vesicle formation
endocrine pancreas development
negative regulation of chondrocyte differentiation
positive regulation of chondrocyte differentiation
lacrimal gland development
protein localization to nucleus
somatic stem cell population maintenance
intrahepatic bile duct development
regulation of cell proliferation
regulation of apoptotic process
negative regulation of apoptotic process
protein kinase B signaling
negative regulation of myoblast differentiation
positive regulation of protein catabolic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of photoreceptor cell differentiation
oligodendrocyte differentiation
positive regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
cartilage development
Sertoli cell differentiation
Sertoli cell development
astrocyte fate commitment
retina development in camera-type eye
limb bud formation
retinal rod cell differentiation
epithelial tube branching involved in lung morphogenesis
lung epithelial cell differentiation
epithelial cell proliferation involved in prostatic bud elongation
bronchus cartilage development
trachea cartilage development
intestinal epithelial structure maintenance
regulation of cell proliferation involved in tissue homeostasis
positive regulation of cartilage development
regulation of branching involved in lung morphogenesis
morphogenesis of a branching epithelium
lung smooth muscle development
negative regulation of biomineral tissue development
ERK1 and ERK2 cascade
Harderian gland development
cellular response to mechanical stimulus
cellular response to retinoic acid
cellular response to interleukin-1
cellular response to epidermal growth factor stimulus
cellular response to heparin
cellular response to transforming growth factor beta stimulus
renal vesicle induction
ureter urothelium development
ureter smooth muscle cell differentiation
ureter morphogenesis
metanephric nephron tubule formation
negative regulation of canonical Wnt signaling pathway
cochlea morphogenesis
positive regulation of kidney development
positive regulation of branching involved in ureteric bud morphogenesis
positive regulation of extracellular matrix assembly
positive regulation of male gonad development
positive regulation of cell proliferation involved in heart morphogenesis
positive regulation of mesenchymal stem cell differentiation
regulation of epithelial cell proliferation involved in lung morphogenesis
negative regulation of mesenchymal cell apoptotic process
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
enhancer sequence-specific DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein kinase activity
protein binding
beta-catenin binding
protein kinase A catalytic subunit binding
enhancer binding
bHLH transcription factor binding
protein heterodimerization activity
pre-mRNA intronic binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-2026.03.11-1.0