Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
APOC3
Transcript:
NM_000040.1
Assay Name:
Mutation genome location:
chr.11 116830554-116830817 on build GRCh38
Assay ID
Hs00834425_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
apolipoprotein
Gene Ontology Categories:
Function(s)
Process(es)
retinoid metabolic process
triglyceride metabolic process
inflammatory response
G-protein coupled receptor signaling pathway
positive regulation of triglyceride biosynthetic process
negative regulation of triglyceride catabolic process
negative regulation of very-low-density lipoprotein particle remodeling
negative regulation of very-low-density lipoprotein particle clearance
negative regulation of high-density lipoprotein particle clearance
negative regulation of low-density lipoprotein particle clearance
triglyceride catabolic process
regulation of Cdc42 protein signal transduction
response to vitamin A
cholesterol efflux
phospholipid efflux
response to triglyceride
high-density lipoprotein particle remodeling
very-low-density lipoprotein particle assembly
chylomicron remnant clearance
lipoprotein metabolic process
response to drug
cholesterol homeostasis
lipoprotein transport
response to peptide hormone
reverse cholesterol transport
negative regulation of fatty acid biosynthetic process
negative regulation of lipid metabolic process
negative regulation of receptor-mediated endocytosis
negative regulation of lipid catabolic process
negative regulation of lipoprotein lipase activity
negative regulation of cholesterol import
triglyceride homeostasis
response to fatty acid
cellular response to glucose stimulus
negative regulation of oxidative phosphorylation
response to simvastatin
phospholipid binding
cholesterol binding
enzyme regulator activity
lipase inhibitor activity
high-density lipoprotein particle receptor binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline