Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00129514_cn
          See other FN1 CNV Assays ›
          Gene Symbol
          FN1
          Assay Reference Genome
          Location

          Chr.2:215378274 on build GRCh38
          Cytoband
          2q35
          Assay ID Hs00129514_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          2335

          Gene Name:

          fibronectin 1

          Gene Aliases:

          CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF

          Location:

          Chr.2:215360440-215436167 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 34 - Exon 35
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          FN1 NM_001306129.1 NP_001293058.1
          NM_001306130.1 NP_001293059.1
          NM_001306131.1 NP_001293060.1
          NM_001306132.1 NP_001293061.1
          NM_002026.3 NP_002017.1
          NM_212474.2 NP_997639.1
          NM_212476.2 NP_997641.1
          NM_212478.2 NP_997643.1
          NM_212482.2 NP_997647.1
          XM_005246397.1 XP_005246454.1
          XM_005246398.1 XP_005246455.1
          XM_005246399.1 XP_005246456.1
          XM_005246401.1 XP_005246458.1
          XM_005246402.1 XP_005246459.1
          XM_005246403.1 XP_005246460.1
          XM_005246404.1 XP_005246461.1
          XM_005246405.1 XP_005246462.1
          XM_005246406.1 XP_005246463.1
          XM_005246407.1 XP_005246464.1
          XM_005246408.1 XP_005246465.1
          XM_005246409.1 XP_005246466.1
          XM_005246410.1 XP_005246467.1
          XM_005246411.1 XP_005246468.1
          XM_005246412.1 XP_005246469.1
          XM_005246414.1 XP_005246471.1
          XM_005246416.1 XP_005246473.1
          XM_017003692.1 XP_016859181.1
          XM_017003693.1 XP_016859182.1
          XM_017003694.1 XP_016859183.1
          XM_017003695.1 XP_016859184.1
          AB191261.1 BAD52437.1
          AB209287.1 BAD92524.1
          AB209840.1 BAD93077.1
          AJ320525.1 CAC86914.1
          AJ320526.1 CAC86915.1
          AJ320527.1 CAC86916.1
          AK300246.1
          AK300458.1
          AL832202.1 CAD91166.1
          AL832771.1
          BC117176.1
          BC143754.1
          BC143763.1
          BX537590.1 CAD97791.1
          BX538017.1 CAD97964.1
          BX538018.1 CAD97965.1
          BX640608.1 CAE45714.1
          BX640638.1 CAE45786.1
          BX640731.1 CAE45847.1
          BX640802.1 CAE45885.1
          BX640803.1
          BX640875.1 CAE45932.1
          BX640920.1 CAE45958.1
          BX640999.1 CAE46002.1
          BX641150.1
          BX649182.1 CAE46200.1
          CR749281.1 CAH18136.1
          CR749316.1 CAH18171.1
          CR749317.1 CAH18172.1
          CR749666.1 CAH18457.1
          CR936623.1 CAI56766.1
          EF550130.1
          EF550131.1
          EF550132.1
          EF550133.1
          EF550134.1
          EF550135.1
          M10905.1 AAA52462.1
          U42458.1 AAD09450.1
          U42593.1 AAD00018.1
          U42594.1 AAD00019.1
          U60067.1 AAD11500.1
          U60068.1 AAD10853.1
          X02761.1 CAA26536.1

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic non-DGV Variation

          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          cell activation
          regulation of protein phosphorylation
          platelet degranulation
          response to ischemia
          acute-phase response
          cell-substrate junction assembly
          cell adhesion
          calcium-independent cell-matrix adhesion
          positive regulation of cell proliferation
          glial cell migration
          regulation of cell shape
          response to wounding
          response to ozone
          positive regulation of gene expression
          positive regulation of peptidase activity
          peptide cross-linking
          extracellular matrix disassembly
          extracellular matrix organization
          integrin activation
          substrate adhesion-dependent cell spreading
          cellular response to vascular endothelial growth factor stimulus
          endodermal cell differentiation
          cellular response to platelet-derived growth factor stimulus
          wound healing
          negative regulation of apoptotic process
          positive regulation of axon extension
          positive regulation of fibroblast proliferation
          leukocyte migration
          positive regulation of chemotaxis
          response to glucocorticoid
          regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          cellular response to mercury ion
          cellular response to glucose stimulus
          cellular response to interleukin-1
          cellular response to prostaglandin E stimulus
          cellular response to transforming growth factor beta stimulus
          cellular response to BMP stimulus
          positive regulation of substrate-dependent cell migration, cell attachment to substrate
          negative regulation of transforming growth factor-beta secretion
          protease binding
          integrin binding
          protein binding
          collagen binding
          heparin binding
          peptidase activator activity
          identical protein binding
          mercury ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline