Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00210178_cn
          See other USP19 CNV Assays ›
          Gene Symbol
          USP19
          Assay Reference Genome
          Location

          Chr.3:49110316 on build GRCh38
          Cytoband
          3p21.31
          Assay ID Hs00210178_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          10869

          Gene Name:

          ubiquitin specific peptidase 19

          Gene Aliases:

          ZMYND9

          Location:

          Chr.3:49108046-49120938 on Build GRCh38

          Assay Gene Location:

          Within Exon 26
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          USP19 NM_001199160.1 26 4219 NP_001186089.1
          NM_001199161.1 26 4225 NP_001186090.1
          NM_001199162.1 26 4189 NP_001186091.1
          NM_006677.2 25 3916 NP_006668.1
          XM_005264823.2 26 4226 XP_005264880.1
          XM_005264824.2 26 4245 XP_005264881.1
          XM_005264825.2 26 4220 XP_005264882.1
          XM_005264826.2 26 4220 XP_005264883.1
          XM_005264827.2 26 4181 XP_005264884.1
          XM_005264829.2 26 4226 XP_005264886.1
          XM_005264830.2 25 3923 XP_005264887.1
          XM_005264831.2 25 3872 XP_005264888.1
          XM_006712946.2 26 4223 XP_006713009.1
          XM_006712947.2 26 4172 XP_006713010.1
          XM_006712948.2 26 4214 XP_006713011.1
          XM_006712949.2 26 4076 XP_006713012.1
          XM_006712950.2 26 4175 XP_006713013.1
          XM_006712951.2 26 4172 XP_006713014.1
          XM_006712952.2 26 4169 XP_006713015.1
          XM_006712953.1 26 4140 XP_006713016.1
          XM_017005615.1 26 4217 XP_016861104.1
          XM_017005616.1 26 4211 XP_016861105.1
          XM_017005617.1 26 4175 XP_016861106.1
          XM_017005618.1 26 4220 XP_016861107.1
          XM_017005619.1 26 4217 XP_016861108.1
          XM_017005620.1 26 4211 XP_016861109.1
          XM_017005621.1 26 4070 XP_016861110.1
          XM_017005622.1 26 4181 XP_016861111.1
          XM_017005623.1 26 4064 XP_016861112.1
          XM_017005624.1 26 4064 XP_016861113.1
          XM_017005625.1 25 3917 XP_016861114.1
          XM_017005626.1 25 3930 XP_016861115.1
          XM_017005627.1 25 3878 XP_016861116.1
          XM_017005628.1 25 3872 XP_016861117.1
          XM_017005629.1 25 3987 XP_016861118.1
          XM_017005630.1 25 3923 XP_016861119.1
          XM_017005631.1 25 3917 XP_016861120.1
          XM_017005632.1 25 3917 XP_016861121.1
          XM_017005633.1 25 3911 XP_016861122.1
          XM_017005634.1 25 3878 XP_016861123.1
          XM_017005635.1 19 2654 XP_016861124.1
          AB020698.1 25 3758
          AK294756.1 26 4112
          AK300425.1 26 4043
          BC048269.1 7 1064 AAH48269.1
          BC065909.1 14 1996 AAH65909.1
          BC082241.1 23 3292 AAH82241.1
          BC106029.1 AAI06030.1
          BC142660.1 26 4095
          BC142727.1 26 4101
          BC146752.1 25 3758
          EF036501.1 3 227

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv834686 Chr.3:49068614 - 49233123 on Build GRCh38 Loss MIR6890 LAMB2P1 CCDC36 USP19 LAMB2 C3orf84 KLHDC8B CCDC71 QRICH1 QARS

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          protein deubiquitination
          ER-associated ubiquitin-dependent protein catabolic process
          regulation of protein stability
          response to endoplasmic reticulum stress
          negative regulation of skeletal muscle tissue development
          protein stabilization
          protein K48-linked deubiquitination
          positive regulation of cell cycle process
          regulation of cellular response to hypoxia
          negative regulation of proteasomal protein catabolic process
          regulation of ERAD pathway
          thiol-dependent ubiquitin-specific protease activity
          protein binding
          ubiquitin protein ligase binding
          metal ion binding
          Hsp90 protein binding
          Lys48-specific deubiquitinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0