Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00224840_cn
          See other DUSP18 CNV Assays ›
          Gene Symbol
          DUSP18
          Assay Reference Genome
          Location

          Chr.22:30663161 on build GRCh38
          Cytoband
          Assay ID Hs00224840_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          150290

          Gene Name:

          dual specificity phosphatase 18

          Gene Aliases:

          DSP18, DUSP20, LMWDSP20

          Location:

          Chr.22:30652051-30667875 on Build GRCh38

          Assay Gene Location:

          Within Exon 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          DUSP18 NM_001304794.2 3 1501 NP_001291723.1
          NM_001304795.2 NP_001291724.1
          NM_001304796.2 3 1224 NP_001291725.1
          NM_152511.5 2 1334 NP_689724.3
          XM_005261368.6 3 1191 XP_005261425.1
          XM_011529921.4 4 1573 XP_011528223.1
          XM_017028627.3 4 1323 XP_016884116.1
          XM_017028628.2 XP_016884117.1
          XM_047441158.1 4 1358 XP_047297114.1
          XM_047441159.1 5 1490 XP_047297115.1
          AF461689.1 2 1344 AAN77931.1
          AK025036.1 1 522
          AK056074.1 2 1344 BAG51616.1
          BC030987.1 AAH30987.1

          Target Gene Details

          Entrez Gene ID:

          339665

          Gene Name:

          solute carrier family 35 member E4

          Gene Aliases:

          -

          Location:

          Chr.22:30635781-30669019 on Build GRCh38

          Assay Gene Location:

          Within Exon 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SLC35E4 NM_001318370.2 3 2485 NP_001305299.1
          NM_001318371.2 NP_001305300.1
          BC093099.1 3 1943 AAH93099.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv834175 Chr.22:30580637 - 30693558 on Build GRCh38 Gain PES1 TCN2 SLC35E4 DUSP18
          nsv6549241 Chr.22:30659456 - 30688376 on Build GRCh38 Duplication SLC35E4 DUSP18

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          dephosphorylation
          peptidyl-tyrosine dephosphorylation
          peptidyl-threonine dephosphorylation
          transmembrane transport
          phosphoprotein phosphatase activity
          protein serine/threonine phosphatase activity
          protein tyrosine phosphatase activity
          protein binding
          protein tyrosine/serine/threonine phosphatase activity
          hydrolase activity
          phosphatase activity
          MAP kinase tyrosine/serine/threonine phosphatase activity
          antiporter activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline