Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00246759_cn
          See other NAPRT CNV Assays ›
          Gene Symbol
          NAPRT
          Assay Reference Genome
          Location

          Chr.8:143575664 on build GRCh38
          Cytoband
          Assay ID Hs00246759_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          93100

          Gene Name:

          nicotinate phosphoribosyltransferase

          Gene Aliases:

          NAPRT1, PP3856

          Location:

          Chr.8:143572874-143578330 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 8 - Intron 9
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          NAPRT NM_001286829.2 NP_001273758.1
          NM_001363145.1 NP_001350074.1
          NM_001363146.1 NP_001350075.1
          NM_145201.6 NP_660202.3
          XM_047422423.1 XP_047278379.1
          AB242230.1 BAF75377.1
          AF258565.1 AAG23768.1
          AK097002.1
          AY214325.1 AAP69603.1
          AY214326.1 AAP69604.1
          AY214327.1 AAP69605.1
          BC006284.2 AAH06284.2
          BC032466.1 AAH32466.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv470247 Chr.8:143283403 - 143615483 on Build GRCh38 Loss RHPN1 SNORD149 GFUS ZC3H3 TIGD5 LOC100310756 MAFA MROH6 NAPRT RHPN1-AS1 GSDMD TOP1MT EEF1D ZNF696 MAFA-AS1 PYCR3
          nsv4171477 Chr.8:143406532 - 143804137 on Build GRCh38 Duplication SNORD149 MAPK15 GFUS ZC3H3 ZNF623 TIGD5 LINC02878 SCRIB LOC100310756 ZNF707 MAFA CCDC166 MROH6 NAPRT LOC105375798 LOC105375800 GSDMD LOC101928160 IQANK1 FAM83H EEF1D MAFA-AS1 PYCR3 MIR4664
          dgv12418n54 Chr.8:143574584 - 143611897 on Build GRCh38 Loss TIGD5 EEF1D NAPRT PYCR3
          nsv1161810 Chr.8:143542268 - 143622987 on Build GRCh38 Deletion LOC105375798 GSDMD GFUS TIGD5 LOC100310756 EEF1D MROH6 NAPRT PYCR3
          nsv612793 Chr.8:143552167 - 143630578 on Build GRCh38 Loss LOC105375798 GSDMD GFUS TIGD5 LOC100310756 EEF1D MROH6 NAPRT PYCR3
          nsv516210 Chr.8:143533025 - 143615483 on Build GRCh38 Loss GSDMD GFUS ZC3H3 TIGD5 LOC100310756 EEF1D MROH6 NAPRT PYCR3
          nsv612774 Chr.8:143489261 - 143606976 on Build GRCh38 Loss SNORD149 GSDMD ZC3H3 TIGD5 LOC100310756 EEF1D MROH6 NAPRT PYCR3
          nsv950013 Chr.8:143465031 - 143616230 on Build GRCh38 Deletion SNORD149 GSDMD GFUS ZC3H3 TIGD5 LOC100310756 EEF1D MROH6 NAPRT PYCR3
          nsv1161809 Chr.8:143467990 - 143610108 on Build GRCh38 Deletion SNORD149 GSDMD ZC3H3 TIGD5 LOC100310756 EEF1D MROH6 NAPRT PYCR3
          nsv470248 Chr.8:143549816 - 143582424 on Build GRCh38 Gain GSDMD LOC100310756 EEF1D MROH6 NAPRT
          nsv612777 Chr.8:143538914 - 143632493 on Build GRCh38 Gain LOC105375798 GSDMD GFUS ZC3H3 TIGD5 LOC100310756 EEF1D MROH6 NAPRT PYCR3
          nsv612812 Chr.8:143563290 - 143592878 on Build GRCh38 Loss EEF1D MROH6 NAPRT
          nsv6425791 Chr.8:142975701 - 144021300 on Build GRCh38 Duplication RHPN1 MAPK15 SPATC1 LINC02878 MAFA MROH6 MIR661 MIR6845 PLEC PARP10 GPIHBP1 ZNF696 PYCR3 LY6E MIR4664 LY6H LY6S ZC3H3 SCRIB NAPRT MINCR NRBP2 LY6E-DT LOC105375798 TOP1MT LY6L ZFP41 CDC42P3 LOC105375800 LOC101928160 GLI4 PUF60 LY6S-AS1 SNORD149 GFUS ZNF623 TIGD5 LOC100310756 MIR937 ZNF707 CCDC166 GRINA RHPN1-AS1 GSDMD IQANK1 FAM83H EPPK1 EEF1D MAFA-AS1
          nsv6435049 Chr.8:143430801 - 143727600 on Build GRCh38 Duplication SNORD149 MAPK15 GFUS ZC3H3 ZNF623 TIGD5 LINC02878 LOC100310756 ZNF707 CCDC166 MROH6 NAPRT LOC105375798 GSDMD LOC101928160 FAM83H EEF1D PYCR3
          dgv7336n100 Chr.8:143510004 - 143616975 on Build GRCh38 Gain GSDMD GFUS ZC3H3 TIGD5 LOC100310756 EEF1D MROH6 NAPRT PYCR3
          esv2738021 Chr.8:143575659 - 143575849 on Build GRCh38 Deletion NAPRT
          nsv824770 Chr.8:143560533 - 143579456 on Build GRCh38 Gain GSDMD MROH6 NAPRT
          nsv612813 Chr.8:143566472 - 143611669 on Build GRCh38 Loss TIGD5 EEF1D MROH6 NAPRT PYCR3
          esv2738020 Chr.8:143575546 - 143576068 on Build GRCh38 Deletion NAPRT
          nsv466000 Chr.8:143551386 - 143582424 on Build GRCh38 Loss GSDMD LOC100310756 EEF1D MROH6 NAPRT

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          response to oxidative stress
          NAD biosynthetic process
          pyridine nucleotide biosynthetic process
          NAD salvage
          nicotinate phosphoribosyltransferase activity
          protein binding
          transferase activity
          ligase activity
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline