Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00278182_cn
          See other PEX6 CNV Assays ›
          Gene Symbol
          PEX6
          Assay Reference Genome
          Location

          Chr.6:42974991 on build GRCh38
          Cytoband
          Assay ID Hs00278182_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5190

          Gene Name:

          peroxisomal biogenesis factor 6

          Gene Aliases:

          HMLR2, PAF-2, PAF2, PBD4A, PDB4B, PXAAA1

          Location:

          Chr.6:42963865-42979181 on Build GRCh38

          Assay Gene Location:

          Within Exon 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PEX6 NM_000287.4 2 961 NP_000278.3
          NM_001316313.2 2 697 NP_001303242.1
          NR_133009.2 2 961
          XM_011514661.3 2 961 XP_011512963.1
          XM_047418872.1 XP_047274828.1
          XM_047418873.1 2 961 XP_047274829.1
          AB051076.1 2 930 BAB83046.1
          AB051077.1 2 666 BAB83047.1
          AB051078.1 2 930 BAB83048.1
          AB208906.1 2 983 BAD92143.1
          AK225704.1 2 961
          AK314237.1 2 959 BAG36906.1
          BC048331.1 2 946 AAH48331.1
          D83703.1 2 1000 BAA12069.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3608809 Chr.6:42973447 - 42975681 on Build GRCh38 Loss PEX6
          esv3576145 Chr.6:42728097 - 43012090 on Build GRCh38 Gain TBCC CNPY3 PPP2R5D BICRAL PTCRA PEX39 GNMT ATP6V0CP3 LOC401261 MEA1 RPL7L1 LINC02976 PEX6 CNPY3-GNMT
          nsv830651 Chr.6:42845563 - 43000871 on Build GRCh38 Loss RPL7L1 LINC02976 CNPY3 PPP2R5D BICRAL PEX6 PTCRA PEX39 CNPY3-GNMT GNMT
          nsv602992 Chr.6:42948182 - 43110718 on Build GRCh38 Loss KLC4 MEA1 CUL7 KLHDC3 LINC02976 MRPL2 PTK7 PPP2R5D PEX6 RRP36 CNPY3-GNMT GNMT
          nsv5899267 Chr.6:42957850 - 43031425 on Build GRCh38 Deletion MEA1 KLHDC3 PPP2R5D PEX6 RRP36 CNPY3-GNMT GNMT
          nsv4147691 Chr.6:42940459 - 42987170 on Build GRCh38 Duplication LINC02976 PPP2R5D PEX6 CNPY3-GNMT GNMT
          nsv1017398 Chr.6:42659705 - 43400998 on Build GRCh38 Gain TTBK1 LOC124901227 CNPY3 BICRAL SLC22A7 PEX39 GNMT ZNF318 KLC4 PRPH2 RPL7L1 KLHDC3 LINC02976 MRPL2 UBR2 PTK7 SRF PEX6 CNPY3-GNMT TBCC CRIP3 CUL9 PPP2R5D PTCRA RRP36 ATP6V0CP3 LOC401261 MEA1 CUL7 DNPH1
          nsv5465493 Chr.6:42973563 - 42975786 on Build GRCh38 Deletion PEX6

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to peroxisome
          peroxisome organization
          protein import into peroxisome matrix
          protein import into peroxisome matrix, translocation
          protein import into peroxisome matrix, receptor recycling
          protein unfolding
          protein stabilization
          nucleotide binding
          protein binding
          ATP binding
          hydrolase activity
          ATPase activity
          macromolecular complex binding
          ubiquitin-dependent protein binding
          protein transporter activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline