Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00315279_cn
          See other NSUN6 CNV Assays ›
          Gene Symbol
          NSUN6
          Assay Reference Genome
          Location

          Chr.10:18616228 on build GRCh38
          Cytoband
          Assay ID Hs00315279_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          221078

          Gene Name:

          NOP2/Sun RNA methyltransferase 6

          Gene Aliases:

          4933414E04Rik, ARL5B-AS1, MRT82, NOPD1

          Location:

          Chr.10:18545561-18659327 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 8 - Exon 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          NSUN6 NM_001351115.2 NP_001338044.1
          NM_001351116.2 NP_001338045.1
          NM_001351117.2 NP_001338046.1
          NM_001351118.2 NP_001338047.1
          NM_182543.5 NP_872349.1
          XM_011519383.2 XP_011517685.1
          XM_011519384.2 XP_011517686.1
          XM_011519385.2 XP_011517687.1
          XM_011519386.3 XP_011517688.1
          XM_011519387.3 XP_011517689.1
          XM_017015907.3 XP_016871396.1
          XM_017015912.3 XP_016871401.1
          XM_024447884.2 XP_024303652.1
          XM_047424777.1 XP_047280733.1
          XM_047424778.1 XP_047280734.1
          XM_047424779.1 XP_047280735.1
          XM_047424780.1 XP_047280736.1
          XM_047424781.1 XP_047280737.1
          XM_047424782.1 XP_047280738.1
          XM_047424783.1 XP_047280739.1
          XM_047424784.1 XP_047280740.1
          AK074323.1 BAB85051.1
          BC035778.1 AAH35778.1
          BQ224447.1
          DA332885.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4744042 Chr.10:18609226 - 18622755 on Build GRCh38 Deletion NSUN6
          esv3622513 Chr.10:18605251 - 18653373 on Build GRCh38 Loss NSUN6
          nsv5475053 Chr.10:18549716 - 18653624 on Build GRCh38 Deletion NSUN6
          nsv1154555 Chr.10:18576029 - 18655757 on Build GRCh38 Deletion NSUN6
          nsv5490847 Chr.10:18541536 - 18621352 on Build GRCh38 Duplication NSUN6 CACNB2
          esv3622511 Chr.10:18550373 - 18655215 on Build GRCh38 Loss NSUN6
          nsv550044 Chr.10:18550421 - 18617006 on Build GRCh38 Loss NSUN6
          esv2759736 Chr.10:18546396 - 18617578 on Build GRCh38 Gain NSUN6
          nsv550042 Chr.10:18394338 - 18705654 on Build GRCh38 Gain NSUN6 CACNB2 ARL5B
          esv3622510 Chr.10:18542215 - 18621057 on Build GRCh38 Gain NSUN6 CACNB2
          esv34545 Chr.10:18532256 - 18619365 on Build GRCh38 Gain NSUN6 CACNB2

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA methylation
          tRNA C5-cytosine methylation
          RNA processing
          tRNA modification
          tRNA methylation
          methylation
          tRNA binding
          RNA binding
          methyltransferase activity
          RNA methyltransferase activity
          S-adenosylmethionine-dependent methyltransferase activity
          tRNA (cytosine-5-)-methyltransferase activity
          transferase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline