Search
Search

Entrez Gene ID: | 23623 |
Gene Name: | RUN and SH3 domain containing 1 |
Gene Aliases: |
NESCA |
Location: |
Chr.1:155320894-155331118 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831636 | Chr.1:155249432 - 155475073 on Build GRCh38 | Loss |
|
| esv33869 | Chr.1:155253492 - 155948170 on Build GRCh38 | Loss |
|
| nsv946413 | Chr.1:155317158 - 155326869 on Build GRCh38 | Duplication |
|
| esv2758971 | Chr.1:155122502 - 155345016 on Build GRCh38 | Gain |
|
Set Membership: |
|