Search
Search

Entrez Gene ID: | 80155 |
Gene Name: | N(alpha)-acetyltransferase 15, NatA auxiliary subunit |
Gene Aliases: |
Ga19, NARG1, NAT1P, NATH, TBDN, TBDN100 |
Location: |
Chr.4:139301467-139390781 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA15 | NM_057175.3 | 20 | 3230 | NP_476516.1 |
| XM_005263236.2 | 20 | 3282 | XP_005263293.1 | |
| AF327722.1 | 20 | 3253 | AAK15707.1 | |
| AJ314788.1 | 20 | 3182 | CAC43228.1 | |
| AK001595.1 | 2 | 681 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830084 | Chr.4:139326103 - 139478732 on Build GRCh38 | Loss |
|
| nsv1023892 | Chr.4:139377986 - 139483575 on Build GRCh38 | Gain |
|
| dgv5409n100 | Chr.4:139326161 - 139396715 on Build GRCh38 | Loss |
|
Set Membership: |
|