Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00505581_cn
          See other CXCL12 CNV Assays ›
          Gene Symbol
          CXCL12
          Assay Reference Genome
          Location

          Chr.10:44371312 on build GRCh38
          Cytoband
          10q11.21
          Assay ID Hs00505581_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          6387

          Gene Name:

          C-X-C motif chemokine ligand 12

          Gene Aliases:

          IRH, PBSF, SCYB12, SDF1, TLSF, TPAR1

          Location:

          Chr.10:44292088-44385097 on Build GRCh38

          Assay Gene Location:

          Within Exon 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CXCL12 NM_000609.6 4 2390 NP_000600.1
          NM_001277990.1 3 1980 NP_001264919.1
          AK124641.1 1 3461
          AL713778.1 1 3250 CAD28539.1
          BX647204.1 1 4192
          DQ345519.1 ABC69272.1
          DQ345520.1 ABC69273.1
          L36033.1 4 2377 AAB39332.1
          U16752.1 4 2378 AAA97434.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv483001 Chr.10:44208843 - 44393301 on Build GRCh38 Loss C10orf142 CXCL12
          nsv1054649 Chr.10:44353473 - 44394265 on Build GRCh38 Gain CXCL12
          nsv1038511 Chr.10:44101437 - 44374582 on Build GRCh38 Loss C10orf142 CXCL12
          nsv825357 Chr.10:44371199 - 44371824 on Build GRCh38 Gain CXCL12

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          neuron migration
          positive regulation of endothelial cell proliferation
          cellular calcium ion homeostasis
          chemotaxis
          immune response
          cell adhesion
          signal transduction
          G-protein coupled receptor signaling pathway
          axon guidance
          blood circulation
          regulation of actin polymerization or depolymerization
          adult locomotory behavior
          response to radiation
          response to heat
          response to mechanical stimulus
          response to virus
          telencephalon cell migration
          organ regeneration
          positive regulation of dopamine secretion
          response to peptide hormone
          positive regulation of cell adhesion
          positive regulation of axon extension involved in axon guidance
          positive chemotaxis
          induction of positive chemotaxis
          cell chemotaxis
          chemokine-mediated signaling pathway
          positive regulation of monocyte chemotaxis
          positive regulation of calcium ion import
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of leukocyte tethering or rolling
          negative regulation of leukocyte apoptotic process
          positive regulation of T cell migration
          receptor binding
          chemokine activity
          growth factor activity
          chemoattractant activity
          chemokine receptor binding
          CXCR chemokine receptor binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline