Search
Search

Entrez Gene ID: | 987 |
Gene Name: | LPS responsive beige-like anchor protein |
Gene Aliases: |
BGL, CDC4L, CVID8, LAB300, LBA, uc.147 |
Location: |
Chr.4:150264435-151015727 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 39 - Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRBA | NM_001199282.3 | NP_001186211.2 | ||
| NM_001364905.1 | NP_001351834.1 | |||
| NM_001367550.1 | NP_001354479.1 | |||
| NM_001440430.1 | NP_001427359.1 | |||
| NM_001440431.1 | NP_001427360.1 | |||
| NM_006726.5 | NP_006717.2 | |||
| XM_005263373.4 | XP_005263430.1 | |||
| XM_011532434.3 | XP_011530736.1 | |||
| XM_047416462.1 | XP_047272418.1 | |||
| AB208838.1 | BAD92075.1 | |||
| AF216648.2 | AAG48558.2 | |||
| AF467287.1 | AAM53530.1 | |||
| AK122920.1 | BAG53798.1 | |||
| M83822.1 | AAB09603.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1024820 | Chr.4:150442926 - 151022409 on Build GRCh38 | Gain |
|
| nsv4547 | Chr.4:150565128 - 150610327 on Build GRCh38 | Deletion |
|
| nsv1032070 | Chr.4:150593512 - 150854107 on Build GRCh38 | Loss |
|
| nsv6386794 | Chr.4:150535901 - 150609700 on Build GRCh38 | Deletion |
|
| nsv4108826 | Chr.4:150484753 - 150615641 on Build GRCh38 | Duplication |
|
Set Membership: |
|