Search
Search

Entrez Gene ID: | 8566 |
Gene Name: | pyridoxal kinase |
Gene Aliases: |
C21orf124, C21orf97, HEL-S-1a, HMSN6C, PKH, PNK, PRED79 |
Location: |
Chr.21:43719129-43762299 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDXK | NM_001331030.2 | NP_001317959.1 | ||
| NM_003681.5 | NP_003672.1 | |||
| XM_005261195.3 | XP_005261252.2 | |||
| XM_005261196.4 | XP_005261253.1 | |||
| XM_005261199.3 | XP_005261256.1 | |||
| XM_011529761.2 | XP_011528063.1 | |||
| XM_047441001.1 | XP_047296957.1 | |||
| XM_047441002.1 | XP_047296958.1 | |||
| XM_047441003.1 | XP_047296959.1 | |||
| XM_047441004.1 | XP_047296960.1 | |||
| XM_047441005.1 | XP_047296961.1 | |||
| XM_047441006.1 | XP_047296962.1 | |||
| AB621819.1 | BAK64155.1 | |||
| AY303972.1 | AAP73047.1 | |||
| BC000123.1 | AAH00123.1 | |||
| BC005825.2 | AAH05825.1 | |||
| BX382033.2 | ||||
| BX382034.2 | ||||
| EU794642.1 | ACJ13696.1 | |||
| U89606.1 | AAC51233.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509799 | Chr.21:43724009 - 43852846 on Build GRCh38 | Insertion |
|
| nsv834107 | Chr.21:43735444 - 43874665 on Build GRCh38 | Loss |
|
| nsv3215180 | Chr.21:43739101 - 43743800 on Build GRCh38 | Deletion |
|
| nsv1072187 | Chr.21:43743018 - 43743719 on Build GRCh38 | Deletion |
|
| nsv470904 | Chr.21:43731164 - 43838166 on Build GRCh38 | Loss |
|
| nsv6056653 | Chr.21:43711677 - 43747683 on Build GRCh38 | Duplication |
|
| nsv587716 | Chr.21:43740206 - 43756740 on Build GRCh38 | Loss |
|
| nsv4273762 | Chr.21:43680575 - 43762549 on Build GRCh38 | Duplication |
|
| dgv7876n54 | Chr.21:43739631 - 43743809 on Build GRCh38 | Loss |
|
| dgv7874n54 | Chr.21:43708309 - 43771258 on Build GRCh38 | Gain |
|
| nsv953642 | Chr.21:43731220 - 43827719 on Build GRCh38 | Deletion |
|
| nsv3234588 | Chr.21:43730270 - 43747789 on Build GRCh38 | Insertion |
|
| esv33771 | Chr.21:43735339 - 43747372 on Build GRCh38 | Gain |
|
Set Membership: |
|