Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00641114_cn
          See other STK38L CNV Assays ›
          Gene Symbol
          STK38L
          Assay Reference Genome
          Location

          Chr.12:27302175 on build GRCh38
          Cytoband
          Assay ID Hs00641114_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          23012

          Gene Name:

          serine/threonine kinase 38 like

          Gene Aliases:

          NDR2

          Location:

          Chr.12:27244286-27325959 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 3 - Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          STK38L NM_015000.4 NP_055815.1
          AB023182.2 BAA76809.3
          AK291055.1 BAF83744.1
          AK304755.1 BAG65510.1
          BC028603.1 AAH28603.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv1406n100 Chr.12:27128236 - 27640822 on Build GRCh38 Gain BMAL2-AS1 BMAL2 STK38L PPFIBP1 SMCO2
          dgv263e214 Chr.12:27152627 - 27304231 on Build GRCh38 Gain STK38L
          nsv6462126 Chr.12:27128016 - 27639839 on Build GRCh38 Duplication BMAL2-AS1 BMAL2 STK38L PPFIBP1 SMCO2
          dgv2417n54 Chr.12:27068022 - 27638988 on Build GRCh38 Gain BMAL2-AS1 BMAL2 C12orf71 STK38L PPFIBP1 SMCO2
          esv3628933 Chr.12:27275994 - 27403117 on Build GRCh38 Gain BMAL2-AS1 BMAL2 STK38L
          nsv4195320 Chr.12:27131567 - 27643067 on Build GRCh38 Deletion BMAL2-AS1 BMAL2 STK38L PPFIBP1 SMCO2
          nsv557808 Chr.12:27288899 - 27339805 on Build GRCh38 Loss BMAL2 STK38L
          dgv271e199 Chr.12:27089700 - 27631451 on Build GRCh38 Deletion BMAL2-AS1 BMAL2 STK38L PPFIBP1 SMCO2
          nsv521872 Chr.12:27114021 - 27635541 on Build GRCh38 Gain BMAL2-AS1 BMAL2 STK38L PPFIBP1 SMCO2
          nsv4528063 Chr.12:27295565 - 27639067 on Build GRCh38 Duplication BMAL2-AS1 BMAL2 STK38L PPFIBP1 SMCO2
          nsv5498428 Chr.12:27130650 - 27643000 on Build GRCh38 Duplication BMAL2-AS1 BMAL2 STK38L PPFIBP1 SMCO2
          nsv436825 Chr.12:27135971 - 27642579 on Build GRCh38 Insertion BMAL2-AS1 BMAL2 STK38L PPFIBP1 SMCO2
          esv2745664 Chr.12:27017516 - 27820117 on Build GRCh38 Deletion BMAL2-AS1 REP15 BMAL2 C12orf71 KLHL42 MANSC4 STK38L MRPS35 MED21 PPFIBP1 SMCO2
          nsv4206867 Chr.12:27109707 - 27308312 on Build GRCh38 Duplication STK38L
          nsv4209820 Chr.12:26484849 - 27683962 on Build GRCh38 Duplication TM7SF3 BMAL2-AS1 BMAL2 C12orf71 FGFR1OP2 STK38L MED21 INTS13 PPFIBP1 ITPR2 SMCO2
          nsv6457231 Chr.12:27127904 - 27639538 on Build GRCh38 Deletion BMAL2-AS1 BMAL2 STK38L PPFIBP1 SMCO2
          nsv5912980 Chr.12:25481034 - 28003055 on Build GRCh38 Deletion BMAL2-AS1 BMAL2 KLHL42 FGFR1OP2 MRPS35 SSPN SSPN-AS1 MIR4302 ITPR2-AS2 ITPR2 RASSF8-AS1 BHLHE41 TM7SF3 C12orf71 MANSC4 STK38L MED21 INTS13 PPFIBP1 LMNTD1 LOC105369705 LOC105369704 REP15 PTHLH RASSF8 SMCO2

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Non-exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          negative regulation of metabolic process
          negative regulation of autophagy
          positive regulation of autophagy
          intracellular signal transduction
          regulation of cellular component organization
          nucleotide binding
          magnesium ion binding
          actin binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          metal ion binding
          protein serine kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline