Search
Search

Entrez Gene ID: | 50515 |
Gene Name: | carbohydrate sulfotransferase 11 |
Gene Aliases: |
C4ST, C4ST-1, C4ST1, HSA269537, OCBMD |
Location: |
Chr.12:104456948-104762014 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHST11 | NM_001173982.2 | 3 | 743 | NP_001167453.1 |
| NM_018413.6 | 3 | 758 | NP_060883.1 | |
| XM_047428914.1 | 2 | 672 | XP_047284870.1 | |
| XM_047428915.1 | 2 | 657 | XP_047284871.1 | |
| AB042326.1 | 3 | 680 | BAA95485.1 | |
| AF239820.1 | 3 | 443 | AAF81691.1 | |
| AJ269537.2 | 3 | 361 | CAB87380.1 | |
| AJ289134.1 | 3 | 419 | CAB92134.1 | |
| AK290923.1 | 3 | 733 | BAF83612.1 | |
| AK313217.1 | 3 | 720 | ||
| BC013315.1 | 4 | 589 | AAH13315.1 |
Set Membership: |
|