Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00894322_cn
          See other GAPDH CNV Assays ›
          Gene Symbol
          GAPDH
          Assay Reference Genome
          Location

          Chr.12:6536816 on build GRCh38
          Cytoband
          Assay ID Hs00894322_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          2597

          Gene Name:

          glyceraldehyde-3-phosphate dehydrogenase

          Gene Aliases:

          G3PD, GAPD, HEL-S-162eP

          Location:

          Chr.12:6534517-6538371 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 4 - Intron 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          GAPDH NM_001256799.3 NP_001243728.1
          NM_001289745.3 NP_001276674.1
          NM_001289746.2 NP_001276675.1
          NM_001357943.2 NP_001344872.1
          NM_002046.7 NP_002037.2
          NR_152150.2
          AB062273.1 BAB93466.1
          AF261085.1 AAF99678.1
          AJ844644.1 CAH59755.1
          AK026525.1
          AK299448.1 BAG61421.1
          AK308198.1
          AY007133.1 AAG01996.1
          AY633612.1 AAV33305.1
          BC001601.1 AAH01601.1
          BC004109.2 AAH04109.1
          BC009081.1 AAH09081.1
          BC013310.2 AAH13310.1
          BC023632.2 AAH23632.1
          BC025925.1 AAH25925.1
          BC026907.1 AAH26907.1
          BC029618.1 AAH29618.1
          BC083511.1 AAH83511.1
          BG724119.1
          BI463134.1
          BM763361.1
          BT006893.1 AAP35539.1
          BU155402.1
          CR407671.1 CAG28599.1
          DQ403057.1 ABD77190.1
          EU668321.1 ACF94474.1
          HY000136.1
          HY004110.1
          HY022295.1
          HY046784.1
          J02642.1 AAA52496.1
          M17851.1 AAA86283.1
          M33197.1 AAA52518.1
          OP866994.1
          X01677.1 CAA25833.1
          X53778.1 CAA37794.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv1357n223 Chr.12:6012501 - 7083700 on Build GRCh38 Duplication ZNF384 VWF ENO2 SCARNA11 MRPL51 RPL13P5 PHB2 LOC105369632 MIR200C USP5 P3H3 SCNN1A CHD4 MIR141 GNB3 EMG1 MLF2 CD9 TAPBPL C1S TNFRSF1A ING4 SCARNA12 SCARNA10 VAMP1 PIANP ACRBP SPSB2 COPS7A CDCA3 CD27-AS1 C12orf57 CHD4-AS1 DSTNP2 IFFO1 NCAPD2 LPAR5 LTBR CD4 LAG3 CD27 LRRC23 PLEKHG6 NOP2 GPR162 ATN1 GAPDH TPI1 MIR200CHG RNU7-1 LPCAT3 PTMS SNORA120 PTPN6 C1R
          nsv8896 Chr.12:6536085 - 6538081 on Build GRCh38 Gain GAPDH
          nsv557255 Chr.12:6531324 - 6538371 on Build GRCh38 Loss GAPDH NCAPD2
          esv2759876 Chr.12:6344718 - 6554669 on Build GRCh38 Gain SCNN1A LTBR CD27-AS1 SCARNA10 GAPDH VAMP1 CD27 MRPL51 IFFO1 TAPBPL NCAPD2
          nsv4205161 Chr.12:6355193 - 6537193 on Build GRCh38 Duplication SCNN1A LTBR CD27-AS1 SCARNA10 GAPDH VAMP1 CD27 MRPL51 TAPBPL NCAPD2
          nsv983266 Chr.12:6536306 - 6538095 on Build GRCh38 Duplication GAPDH

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          positive regulation of cytokine production
          immune system process
          glucose metabolic process
          glycolytic process
          regulation of translation
          apoptotic process
          negative regulation of endopeptidase activity
          regulation of macroautophagy
          negative regulation of translation
          killing of cells of other organism
          positive regulation of type I interferon production
          peptidyl-cysteine S-trans-nitrosylation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          protein stabilization
          defense response to fungus
          neuron apoptotic process
          killing by host of symbiont cells
          antimicrobial humoral immune response mediated by antimicrobial peptide
          cellular response to interferon-gamma
          glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
          protein binding
          microtubule binding
          oxidoreductase activity
          oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
          transferase activity
          aspartic-type endopeptidase inhibitor activity
          peptidyl-cysteine S-nitrosylase activity
          identical protein binding
          NADP binding
          NAD binding
          disordered domain specific binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline