Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00939814_cn
          See other CRIP2 CNV Assays ›
          Gene Symbol
          CRIP2
          Assay Reference Genome
          Location

          Chr.14:105479026 on build GRCh38
          Cytoband
          14q32.33
          Assay ID Hs00939814_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1397

          Gene Name:

          cysteine rich protein 2

          Gene Aliases:

          CRIP, CRP2, ESP1

          Location:

          Chr.14:105472938-105480170 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 7 - Intron 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CRIP2 NM_001270837.1 NP_001257766.1
          NM_001270841.1 NP_001257770.1
          NM_001312.3 NP_001303.1
          NR_073081.1
          NR_073082.1
          NR_073083.1
          NR_073084.1
          NR_073085.1
          AK057837.1
          AK091845.1
          AK223251.1 BAD96971.1
          AK300092.1
          AK311402.1
          AK315757.1
          BC000434.2 AAH00434.1
          BC001931.1 AAH01931.1
          BC034151.1 AAH34151.1
          BC128101.1
          BE271979.1
          BT019911.1 AAV38714.1
          CN367966.1
          D42123.1 BAA07703.1
          HY078632.1
          HY161884.1
          U36190.1 AAB03194.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv3996n54 Chr.14:105474226 - 105488519 on Build GRCh38 Gain CRIP2 CRIP1
          nsv566204 Chr.14:105376842 - 105479048 on Build GRCh38 Gain CRIP2 TEX22 PACS2 LOC100507437 MTA1
          nsv817647 Chr.14:105382554 - 105480505 on Build GRCh38 Gain CRIP2 TEX22 PACS2 LOC100507437 MTA1
          dgv3998n54 Chr.14:105474495 - 105770768 on Build GRCh38 Loss LOC107987211 TMEM121 ELK2AP IGHG1 IGHA1 IGHG3 IGH IGHG2 MIR8071-2 CRIP2 C14orf80 IGHA2 IGHG4 LOC105370697 CRIP1 MIR8071-1
          nsv566229 Chr.14:105474172 - 105483783 on Build GRCh38 Loss CRIP2
          nsv1160204 Chr.14:105291411 - 105483783 on Build GRCh38 Duplication BRF1 CRIP2 TEX22 PACS2 LOC100507437 MTA1
          nsv566206 Chr.14:105439736 - 105543235 on Build GRCh38 Loss CRIP2 C14orf80 TMEM121 LOC105370697 CRIP1 MTA1
          nsv832890 Chr.14:105304955 - 105483089 on Build GRCh38 Loss BRF1 CRIP2 TEX22 PACS2 LOC100507437 MTA1
          nsv1440 Chr.14:105473170 - 105479862 on Build GRCh38 Insertion CRIP2
          esv1004211 Chr.14:105477270 - 105479324 on Build GRCh38 Insertion CRIP2
          nsv566210 Chr.14:105465248 - 105488368 on Build GRCh38 Gain CRIP2 CRIP1 MTA1
          nsv9177 Chr.14:105476516 - 105485998 on Build GRCh38 Gain CRIP2
          nsv827088 Chr.14:105454469 - 105498549 on Build GRCh38 Gain CRIP2 C14orf80 CRIP1 MTA1
          nsv952282 Chr.14:105363764 - 105510163 on Build GRCh38 Deletion CRIP2 C14orf80 TEX22 CRIP1 PACS2 LOC100507437 MTA1
          dgv3999n54 Chr.14:105474867 - 105487998 on Build GRCh38 Loss CRIP2 CRIP1
          esv2760014 Chr.14:105047009 - 105483089 on Build GRCh38 Loss GPR132 BRF1 CRIP2 JAG2 BTBD6 TEX22 PACS2 LOC100507437 MIR6765 MTA1 NUDT14 LOC102723354
          dgv3987n54 Chr.14:105461599 - 105499970 on Build GRCh38 Loss CRIP2 C14orf80 CRIP1 MTA1
          nsv470679 Chr.14:105124547 - 105605978 on Build GRCh38 Loss JAG2 TMEM121 BTBD6 TEX22 IGH MTA1 NUDT14 BRF1 CRIP2 C14orf80 IGHA2 LOC105370697 CRIP1 PACS2 LOC100507437 MIR6765
          nsv566265 Chr.14:105474766 - 105479048 on Build GRCh38 Loss CRIP2
          esv3635645 Chr.14:105473682 - 105484540 on Build GRCh38 Gain CRIP2
          nsv1054190 Chr.14:105246882 - 105701138 on Build GRCh38 Gain TMEM121 BTBD6 TEX22 ELK2AP IGH MTA1 IGHG2 MIR8071-2 BRF1 CRIP2 C14orf80 IGHA2 IGHG4 LOC105370697 CRIP1 PACS2 LOC100507437 MIR8071-1
          dgv3991n54 Chr.14:105472502 - 105480505 on Build GRCh38 Loss CRIP2
          nsv517055 Chr.14:105136282 - 106779068 on Build GRCh38 Gain+Loss BTBD6 MIR4538 IGHD IGHG3 IGH KIAA0125 MTA1 ADAM6 LINC00221 MIR8071-2 CRIP2 IGHV1-69 IGHG4 LOC105370697 MIR4539 MIR6765 IGHV3-71 LOC107987211 IGHV3-41 JAG2 IGHV3-30 TMEM121 TEX22 MIR4537 ELK2AP IGHG1 IGHA1 IGHV3-69-1 IGHV3-23 NUDT14 IGHG2 IGHM BRF1 IGHV4-55 IGHV4-31 C14orf80 IGHA2 MIR4507 CRIP1 PACS2 LINC00226 IGHV3-48 LOC100507437 MIR8071-1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          hemopoiesis
          zinc ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline