Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01193337_cn
          See other PDXK CNV Assays ›
          Gene Symbol
          PDXK
          Assay Reference Genome
          Location

          Chr.21:43759681 on build GRCh38
          Cytoband
          Assay ID Hs01193337_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          8566

          Gene Name:

          pyridoxal kinase

          Gene Aliases:

          C21orf124, C21orf97, HEL-S-1a, HMSN6C, PKH, PNK, PRED79

          Location:

          Chr.21:43719129-43762299 on Build GRCh38

          Assay Gene Location:

          Within Exon 19
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PDXK NM_001331030.2 10 4502 NP_001317959.1
          NM_003681.5 11 4723 NP_003672.1
          XM_005261195.3 11 4907 XP_005261252.2
          XM_005261196.4 11 4599 XP_005261253.1
          XM_005261199.3 11 4488 XP_005261256.1
          XM_011529761.2 10 4466 XP_011528063.1
          XM_047441001.1 12 5436 XP_047296957.1
          XM_047441002.1 10 4525 XP_047296958.1
          XM_047441003.1 9 4471 XP_047296959.1
          XM_047441004.1 12 7518 XP_047296960.1
          XM_047441005.1 12 7660 XP_047296961.1
          XM_047441006.1 11 4488 XP_047296962.1
          AK027018.1
          AL137632.1 1 653
          AL161960.1 1 2299
          DA116202.1 1 316
          DA736443.1 1 49
          EU794642.1 11 4755 ACJ13696.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv509799 Chr.21:43724009 - 43852846 on Build GRCh38 Insertion AATBC CSTB RRP1 PDXK
          nsv834108 Chr.21:43758156 - 43949760 on Build GRCh38 Loss AATBC CSTB RRP1 PDXK AGPAT3
          nsv4273762 Chr.21:43680575 - 43762549 on Build GRCh38 Duplication PDXK RRP1B
          dgv7874n54 Chr.21:43708309 - 43771258 on Build GRCh38 Gain PDXK
          nsv953642 Chr.21:43731220 - 43827719 on Build GRCh38 Deletion AATBC CSTB RRP1 PDXK
          nsv834107 Chr.21:43735444 - 43874665 on Build GRCh38 Loss AATBC CSTB RRP1 PDXK AGPAT3
          nsv470904 Chr.21:43731164 - 43838166 on Build GRCh38 Loss AATBC CSTB RRP1 PDXK
          esv33817 Chr.21:43749531 - 44647625 on Build GRCh38 Loss KRTAP10-1 TRPM2-AS TSPEAR KRTAP10-2 KRTAP10-11 KRTAP10-3 KRTAP10-8 KRTAP10-6 CFAP410 PDXK LOC105372832 LINC02575 AGPAT3 KRTAP10-9 LINC01678 DNMT3L AATBC KRTAP10-4 CSTB KRTAP10-5 DNMT3L-AS1 TSPEAR-AS2 GATD3 LRRC3 AIRE TRAPPC10 PWP2 TRPM2 ICOSLG KRTAP10-10 RRP1 LRRC3-DT PFKL TSPEAR-AS1 KRTAP10-7
          nsv520305 Chr.21:43756077 - 43826445 on Build GRCh38 Loss AATBC CSTB RRP1 PDXK

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          pyridoxal 5'-phosphate salvage
          pyridoxal metabolic process
          pyridoxamine metabolic process
          pyridoxal phosphate metabolic process
          nucleotide binding
          magnesium ion binding
          ATP binding
          zinc ion binding
          pyridoxal kinase activity
          kinase activity
          transferase activity
          phosphotransferase activity, alcohol group as acceptor
          pyridoxal phosphate binding
          potassium ion binding
          sodium ion binding
          lithium ion binding
          protein homodimerization activity
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline