Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01251671_cn
          See other NAPA CNV Assays ›
          Gene Symbol
          NAPA
          Assay Reference Genome
          Location

          Chr.19:47495568 on build GRCh38
          Cytoband
          Assay ID Hs01251671_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          8775

          Gene Name:

          NSF attachment protein alpha

          Gene Aliases:

          SNAPA

          Location:

          Chr.19:47484530-47515063 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 4 - Intron 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          NAPA NM_003827.4 NP_003818.2
          NR_038456.2
          NR_038457.2
          XM_011527436.2 XP_011525738.1
          AK098401.1
          AK126519.1
          AK292244.1 BAF84933.1
          AK308633.1
          AK310114.1
          BC001165.1 AAH01165.1
          BC007432.2 AAH07432.1
          BC028234.1 AAH28234.1
          BC091511.1 AAH91511.1
          BQ948081.1
          BT019568.1 AAV38375.1
          CX873074.1
          U39412.1 AAC80170.1

          Target Gene Details

          Entrez Gene ID:

          100505681

          Gene Name:

          NAPA antisense RNA 1

          Gene Aliases:

          -

          Location:

          Chr.19:47484282-47501597 on Build GRCh38

          Assay Gene Location:

          Within Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          NAPA-AS1 NR_038452.1
          AK057858.1 3 939
          BG035979.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv428366 Chr.19:47354499 - 47522248 on Build GRCh38 Loss MEIS3 ZNF541 KPTN DHX34 SLC8A2 NAPA NAPA-AS1
          nsv1056887 Chr.19:47481963 - 47527369 on Build GRCh38 Gain ZNF541 KPTN NAPA NAPA-AS1
          nsv579888 Chr.19:47428355 - 47658874 on Build GRCh38 Gain BICRA-AS2 ZNF541 KPTN SLC8A2 NAPA BICRA NAPA-AS1
          nsv953591 Chr.19:47486744 - 47500943 on Build GRCh38 Deletion NAPA NAPA-AS1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          intra-Golgi vesicle-mediated transport
          brain development
          regulation of synaptic vesicle priming
          protein transport
          synaptic vesicle priming
          vesicle-mediated transport
          neuron differentiation
          protein-containing complex disassembly
          synaptic transmission, glutamatergic
          SNARE complex disassembly
          apical protein localization
          membrane fusion
          SNARE binding
          soluble NSF attachment protein activity
          protein binding
          syntaxin binding
          macromolecular complex binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline