Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01279572_cn
          See other OR51E1 CNV Assays ›
          Gene Symbol
          OR51E1
          Assay Reference Genome
          Location

          Chr.11:4653392 on build GRCh38
          Cytoband
          Assay ID Hs01279572_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          143503

          Gene Name:

          olfactory receptor family 51 subfamily E member 1

          Gene Aliases:

          D-GPCR, DGPCR, GPR136, GPR164, OR51E1P, OR52A3P, POGR, PSGR2

          Location:

          Chr.11:4643982-4655488 on Build GRCh38

          Assay Gene Location:

          Within Exon 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          OR51E1 NM_152430.4 2 954 NP_689643.2
          AK023643.1
          AK293091.1 2 1517 BAF85780.1
          AL833127.1 2 955
          AY698056.1 2 954 AAU07996.1
          AY775731.1 1 1063 AAV54110.1
          BC022401.1 2 1010 AAH22401.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6442818 Chr.11:4560601 - 4670000 on Build GRCh38 Deletion OR52I2 TRIM68 OR51D1 OR51E1 OR52I1 C11orf40
          nsv6438415 Chr.11:4449872 - 4656651 on Build GRCh38 Deletion OR52I2 TRIM68 OR52K3P OR52M1 OR51D1 OR52K1 OR52K2 OR51E1 OR52I1 C11orf40
          nsv1054705 Chr.11:4560920 - 4666008 on Build GRCh38 Loss OR52I2 TRIM68 OR51D1 OR51E1 OR52I1 C11orf40
          nsv1041633 Chr.11:4618062 - 4671879 on Build GRCh38 Gain OR51D1 OR51E1
          esv3891914 Chr.11:4642313 - 4974456 on Build GRCh38 Gain OR51T1 MMP26 OR51D1 OR51F1 OR51E1 OR51A2 OR51F2 OR51H1 OR51G1 OR52R1 OR51A4 OR51C1 OR51E2 OR51G2 OR51A7 OR51S1
          nsv1054847 Chr.11:4621645 - 5179426 on Build GRCh38 Gain OR51T1 MMP26 OR52E2 OR51L1 OR51D1 OR51F1 OR52J3 OR51E1 OR52A4P OR51A2 OR52Z1P OR51F2 OR51H1 OR52E1 OR51G1 OR52R1 OR52A1 OR51A4 OR52A5 OR51C1 OR51E2 OR51G2 OR51A7 OR51S1
          dgv1586n54 Chr.11:4566710 - 4668512 on Build GRCh38 Loss OR52I2 TRIM68 OR51D1 OR51E1 OR52I1 C11orf40
          nsv4192783 Chr.11:4619101 - 5268941 on Build GRCh38 Duplication OR51T1 HBB OR52J3 OR51E1 OR52A4P OR52Z1P OR51F2 OR52A1 HBG1 OR52A5 OR51C1 OR51V1 BGLT3 HBE1 MMP26 OR52E2 OR51L1 OR51D1 OR51F1 OR51A2 OR51H1 OR52E1 OR51G1 OR52R1 OR51A4 HBD OR51E2 HBBP1 OR51G2 HBG2 OR51A7 OR51S1
          dgv958n223 Chr.11:4619075 - 5268953 on Build GRCh38 Duplication OR51T1 HBB OR52J3 OR51E1 OR52A4P OR52Z1P OR51F2 OR52A1 HBG1 OR52A5 OR51C1 OR51V1 BGLT3 HBE1 MMP26 OR52E2 OR51L1 OR51D1 OR51F1 OR51A2 OR51H1 OR52E1 OR51G1 OR52R1 OR51A4 HBD OR51E2 HBBP1 OR51G2 HBG2 OR51A7 OR51S1
          nsv832056 Chr.11:4651771 - 4803660 on Build GRCh38 Gain OR52R1 MMP26 OR51C1 OR51E2 OR51F1 OR51E1
          nsv832055 Chr.11:4529664 - 4715110 on Build GRCh38 Loss OR52I2 MMP26 OR51C1 TRIM68 OR51E2 OR52M1 OR51D1 OR51E1 OR52I1 C11orf40
          nsv4201396 Chr.11:4560919 - 4669991 on Build GRCh38 Deletion OR52I2 TRIM68 OR51D1 OR51E1 OR52I1 C11orf40

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          system process
          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          cellular response to lipid
          G-protein coupled receptor activity
          olfactory receptor activity
          protein binding
          signaling receptor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline