Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01279866_cn
          See other NSUN6 CNV Assays ›
          Gene Symbol
          NSUN6
          Assay Reference Genome
          Location

          Chr.10:18651175 on build GRCh38
          Cytoband
          Assay ID Hs01279866_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          221078

          Gene Name:

          NOP2/Sun RNA methyltransferase 6

          Gene Aliases:

          4933414E04Rik, ARL5B-AS1, MRT82, NOPD1

          Location:

          Chr.10:18545561-18659327 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 3 - Exon 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          NSUN6 NM_001351115.2 NP_001338044.1
          NM_001351116.2 NP_001338045.1
          NM_001351117.2 NP_001338046.1
          NM_001351118.2 NP_001338047.1
          NM_182543.5 NP_872349.1
          XM_011519383.2 XP_011517685.1
          XM_011519384.2 XP_011517686.1
          XM_011519385.2 XP_011517687.1
          XM_011519386.3 XP_011517688.1
          XM_011519387.3 XP_011517689.1
          XM_017015907.3 XP_016871396.1
          XM_024447884.2 XP_024303652.1
          XM_047424777.1 XP_047280733.1
          XM_047424778.1 XP_047280734.1
          XM_047424779.1 XP_047280735.1
          XM_047424780.1 XP_047280736.1
          XM_047424782.1 XP_047280738.1
          XM_047424784.1 XP_047280740.1
          AK074323.1 BAB85051.1
          BC035778.1 AAH35778.1
          BQ224447.1
          DA332885.1
          DB012566.1
          HY169016.1 1 467

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3622513 Chr.10:18605251 - 18653373 on Build GRCh38 Loss NSUN6
          nsv5475053 Chr.10:18549716 - 18653624 on Build GRCh38 Deletion NSUN6
          nsv1154555 Chr.10:18576029 - 18655757 on Build GRCh38 Deletion NSUN6
          esv3622511 Chr.10:18550373 - 18655215 on Build GRCh38 Loss NSUN6
          nsv6443757 Chr.10:18647752 - 18704055 on Build GRCh38 Deletion NSUN6 ARL5B
          nsv4184187 Chr.10:18648894 - 18656677 on Build GRCh38 Deletion NSUN6
          nsv550042 Chr.10:18394338 - 18705654 on Build GRCh38 Gain NSUN6 ARL5B CACNB2
          nsv466762 Chr.10:18643243 - 18687730 on Build GRCh38 Gain NSUN6 ARL5B
          nsv6015319 Chr.10:18646593 - 18656289 on Build GRCh38 Duplication NSUN6

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA methylation
          tRNA C5-cytosine methylation
          RNA processing
          tRNA modification
          tRNA methylation
          methylation
          tRNA binding
          RNA binding
          methyltransferase activity
          RNA methyltransferase activity
          S-adenosylmethionine-dependent methyltransferase activity
          tRNA (cytosine-5-)-methyltransferase activity
          transferase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline