Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01319300_cn
          See other CNTROB CNV Assays ›
          Gene Symbol
          CNTROB
          Assay Reference Genome
          Location

          Chr.17:7944215 on build GRCh38
          Cytoband
          Assay ID Hs01319300_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          116840

          Gene Name:

          centrobin, centriole duplication and spindle assembly protein

          Gene Aliases:

          LIP8, PP1221

          Location:

          Chr.17:7932081-7949920 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 12 - Intron 12
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CNTROB NM_001037144.7 NP_001032221.1
          NM_001330124.3 NP_001317053.1
          NM_001353202.2 NP_001340131.1
          NM_001353203.2 NP_001340132.1
          NM_001353204.2 NP_001340133.1
          NM_001353205.2 NP_001340134.1
          NM_001353206.2 NP_001340135.1
          NM_001353207.1 NP_001340136.1
          NM_001353208.2 NP_001340137.1
          NM_001353209.1 NP_001340138.1
          NM_053051.5 NP_444279.2
          XM_005256438.4 XP_005256495.1
          XM_017024128.2 XP_016879617.1
          XM_017024129.2 XP_016879618.1
          XM_017024134.3 XP_016879623.1
          XM_017024135.2 XP_016879624.1
          XM_017024137.3 XP_016879626.1
          XM_017024138.2 XP_016879627.1
          XM_017024141.2 XP_016879630.1
          XM_017024143.2 XP_016879632.1
          XM_047435302.1 XP_047291258.1
          XM_047435303.1 XP_047291259.1
          XM_047435304.1 XP_047291260.1
          XM_047435305.1 XP_047291261.1
          AF331638.2 AAL56068.2
          AK074847.1 BAC11241.1
          AK293076.1 BAF85765.1
          AK303093.1 5 671 BAG64203.1
          AL833907.1 CAD38763.1
          AY160226.1 AAO22135.1
          AY160227.1 AAO22136.1
          BC014055.2 AAH14055.3
          BC021134.1 AAH21134.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6500826 Chr.17:7924254 - 7955483 on Build GRCh38 Duplication KCNAB3 CNTROB TRAPPC1
          nsv574333 Chr.17:7880977 - 7952801 on Build GRCh38 Loss KCNAB3 CNTROB RNF227 SCARNA21 TRAPPC1 CHD3 NAA38

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          centriole replication
          centrosome separation
          cell division
          regulation of cilium assembly
          mitotic cytokinetic process
          protein binding
          protein domain specific binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline