Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01331656_cn
          See other TRHDE CNV Assays ›
          Gene Symbol
          TRHDE
          Assay Reference Genome
          Location

          Chr.12:72272394 on build GRCh38
          Cytoband
          Assay ID Hs01331656_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          29953

          Gene Name:

          thyrotropin releasing hormone degrading enzyme

          Gene Aliases:

          PAP-II, PGPEP2, TRH-DE

          Location:

          Chr.12:72087266-72670758 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 2 - Exon 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TRHDE NM_013381.3 NP_037513.2
          XM_005268819.6 XP_005268876.3
          XM_017019243.3 XP_016874732.3
          XM_017019244.2 XP_016874733.1

          Target Gene Details

          Entrez Gene ID:

          283392

          Gene Name:

          TRHDE antisense RNA 1

          Gene Aliases:

          -

          Location:

          Chr.12:72253507-72273509 on Build GRCh38

          Assay Gene Location:

          Within Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TRHDE-AS1 NR_026836.1
          NR_026837.1 1 1116
          AK095156.1 1 133 BAG52994.1
          DA139014.1 1 172

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv832452 Chr.12:72191191 - 72370715 on Build GRCh38 Gain TRHDE TRHDE-AS1
          nsv1044083 Chr.12:72253860 - 72881746 on Build GRCh38 Gain TRHDE TRHDE-AS1
          nsv559362 Chr.12:72248004 - 72300911 on Build GRCh38 Loss TRHDE TRHDE-AS1
          esv2422452 Chr.12:72092153 - 72781801 on Build GRCh38 Duplication TRHDE TRHDE-AS1
          nsv559363 Chr.12:72262145 - 72865334 on Build GRCh38 Gain TRHDE TRHDE-AS1

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          signal transduction
          cell-cell signaling
          peptide catabolic process
          aminopeptidase activity
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          pyroglutamyl-peptidase activity
          metal ion binding
          metalloaminopeptidase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline