Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01426676_cn
          See other CRIP2 CNV Assays ›
          Gene Symbol
          CRIP2
          Assay Reference Genome
          Location

          Chr.14:105478298 on build GRCh38
          Cytoband
          14q32.33
          Assay ID Hs01426676_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1397

          Gene Name:

          cysteine rich protein 2

          Gene Aliases:

          CRIP, CRP2, ESP1

          Location:

          Chr.14:105472938-105480170 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 4 - Exon 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CRIP2 NM_001270837.1 NP_001257766.1
          NM_001270841.1 NP_001257770.1
          NM_001312.3 NP_001303.1
          NR_073081.1
          NR_073082.1
          NR_073083.1
          NR_073084.1
          NR_073085.1
          AK057837.1
          AK091845.1
          AK223251.1 BAD96971.1
          AK300092.1
          AK311402.1
          AK315757.1
          BC000434.2 AAH00434.1
          BC001931.1 AAH01931.1
          BC034151.1 AAH34151.1
          BC128101.1
          BE271979.1
          BT019911.1 AAV38714.1
          CN367966.1
          D42123.1 BAA07703.1
          HY078632.1
          HY161884.1
          U36190.1 AAB03194.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv3996n54 Chr.14:105474226 - 105488519 on Build GRCh38 Gain CRIP2 CRIP1
          nsv566204 Chr.14:105376842 - 105479048 on Build GRCh38 Gain TEX22 LOC100507437 CRIP2 MTA1 PACS2
          nsv817647 Chr.14:105382554 - 105480505 on Build GRCh38 Gain TEX22 LOC100507437 CRIP2 MTA1 PACS2
          dgv3998n54 Chr.14:105474495 - 105770768 on Build GRCh38 Loss IGHG3 IGHG1 MIR8071-2 ELK2AP MIR8071-1 IGHG2 IGHG4 LOC105370697 IGH TMEM121 C14orf80 CRIP2 IGHA2 CRIP1 IGHA1 LOC107987211
          nsv1160204 Chr.14:105291411 - 105483783 on Build GRCh38 Duplication TEX22 LOC100507437 CRIP2 BRF1 MTA1 PACS2
          nsv566229 Chr.14:105474172 - 105483783 on Build GRCh38 Loss CRIP2
          nsv566206 Chr.14:105439736 - 105543235 on Build GRCh38 Loss TMEM121 C14orf80 CRIP2 CRIP1 MTA1 LOC105370697
          nsv832890 Chr.14:105304955 - 105483089 on Build GRCh38 Loss TEX22 LOC100507437 CRIP2 BRF1 MTA1 PACS2
          nsv1440 Chr.14:105473170 - 105479862 on Build GRCh38 Insertion CRIP2
          esv1004211 Chr.14:105477270 - 105479324 on Build GRCh38 Insertion CRIP2
          nsv566210 Chr.14:105465248 - 105488368 on Build GRCh38 Gain CRIP2 CRIP1 MTA1
          nsv9177 Chr.14:105476516 - 105485998 on Build GRCh38 Gain CRIP2
          nsv827088 Chr.14:105454469 - 105498549 on Build GRCh38 Gain C14orf80 CRIP2 CRIP1 MTA1
          nsv952282 Chr.14:105363764 - 105510163 on Build GRCh38 Deletion TEX22 C14orf80 LOC100507437 CRIP2 CRIP1 MTA1 PACS2
          dgv3999n54 Chr.14:105474867 - 105487998 on Build GRCh38 Loss CRIP2 CRIP1
          esv2760014 Chr.14:105047009 - 105483089 on Build GRCh38 Loss BTBD6 TEX22 JAG2 LOC100507437 LOC102723354 GPR132 CRIP2 BRF1 MTA1 PACS2 MIR6765 NUDT14
          nsv470679 Chr.14:105124547 - 105605978 on Build GRCh38 Loss LOC105370697 PACS2 NUDT14 BTBD6 TEX22 IGH TMEM121 JAG2 C14orf80 LOC100507437 CRIP2 IGHA2 BRF1 CRIP1 MTA1 MIR6765
          dgv3987n54 Chr.14:105461599 - 105499970 on Build GRCh38 Loss C14orf80 CRIP2 CRIP1 MTA1
          nsv566265 Chr.14:105474766 - 105479048 on Build GRCh38 Loss CRIP2
          nsv1054190 Chr.14:105246882 - 105701138 on Build GRCh38 Gain MIR8071-2 ELK2AP MIR8071-1 IGHG2 IGHG4 LOC105370697 PACS2 BTBD6 TEX22 IGH TMEM121 C14orf80 LOC100507437 CRIP2 IGHA2 BRF1 CRIP1 MTA1
          esv3635645 Chr.14:105473682 - 105484540 on Build GRCh38 Gain CRIP2
          nsv517055 Chr.14:105136282 - 106779068 on Build GRCh38 Gain+Loss IGHG3 IGHG1 IGHV3-71 ELK2AP IGHG2 IGHG4 MIR4507 LOC105370697 PACS2 KIAA0125 BTBD6 TEX22 MIR4537 IGHM C14orf80 MIR4538 CRIP1 MTA1 IGHA1 LINC00226 MIR6765 MIR8071-2 MIR8071-1 IGHV3-69-1 ADAM6 LINC00221 IGHV3-23 MIR4539 IGHV4-31 NUDT14 IGHV3-48 IGHV3-30 IGH TMEM121 JAG2 LOC100507437 IGHV4-55 CRIP2 IGHA2 BRF1 IGHD IGHV3-41 LOC107987211 IGHV1-69
          dgv3991n54 Chr.14:105472502 - 105480505 on Build GRCh38 Loss CRIP2

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          hemopoiesis
          zinc ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline