Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Entrez Gene ID: | 54982 |
Gene Name: | ceroid-lipofuscinosis, neuronal 6, late infantile, variant |
Gene Aliases: |
CLN4A, HsT18960, nclf |
Location: |
Chr.15:68206992-68229742 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLN6 | NM_017882.2 | 7 | 962 | NP_060352.1 |
AK000568.1 | 7 | 907 | BAA91260.1 | |
AK027604.1 | 7 | 930 | BAB55226.1 | |
AK074753.1 | 4 | 456 | BAC11181.1 | |
AK291175.1 | 7 | 889 | ||
AK293155.1 | 5 | 606 | ||
AK293197.1 | 7 | 1138 | ||
BC010849.1 | 7 | 876 | AAH10849.1 | |
BC013130.1 | 7 | 813 | AAH13130.1 | |
CR457244.1 | 7 | 804 | CAG33525.1 | |
KU178671.1 | 7 | 804 | ||
KU178672.1 | 7 | 923 | ||
KU178673.1 | 6 | 705 | ||
KU178674.1 | 6 | 615 |
Set Membership: |
Intragenic Exonic non-DGV Variation |