Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01495274_cn
          See other TRIM41 CNV Assays ›
          Gene Symbol
          TRIM41
          Assay Reference Genome
          Location

          Chr.5:181224541 on build GRCh38
          Cytoband
          5q35.3
          Assay ID Hs01495274_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          90933

          Gene Name:

          tripartite motif containing 41

          Gene Aliases:

          RINCK

          Location:

          Chr.5:181223263-181235808 on Build GRCh38

          Assay Gene Location:

          Within Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TRIM41 NM_033549.4 1 1279 NP_291027.3
          NM_201627.2 1 1279 NP_963921.1
          NR_045218.1 1 1279
          XM_006714929.2 1 1277 XP_006714992.1
          XM_006714930.2 1 1277 XP_006714993.1
          XM_006714931.3 1 1277 XP_006714994.1
          XM_006714932.3 1 1277 XP_006714995.1
          AB063180.1 1 305 BAB70617.1
          AB100366.1 1 542 BAD07472.1
          AB100367.1 1 542 BAD07473.1
          AK027601.1 1 1252
          AK074052.1 1 376 BAB84878.1
          AL832960.1 1 156 CAH56279.1
          AX775789.1 CAE11594.1
          AX775791.1 CAE11595.1
          AX775793.1 1 1252 CAE11596.1
          AX775797.1 1 310 CAE11598.1
          AX775799.1 1 1252 CAE11599.1
          AX775801.1 1 1252 CAE11600.1
          AX775803.1 1 310 CAE11601.1
          BC004956.1 1 484 AAH04956.1
          BC018765.1 1 310 AAH18765.2
          BC071887.1 1 484 AAH71887.1
          BC090953.1 1 820 AAH90953.1
          BC127195.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv428133 Chr.5:180805451 - 181359487 on Build GRCh38 Gain OR2V2 OR2V1 OR4F3 BTNL8 LOC102577426 TRIM52 CTC-338M12.4 HEIH RACK1 MGAT1 TRIM41 MIR8089 TRIM52-AS1 BTNL3 LOC100132062 SNORD95 BTNL9 TRIM7 ZFP62 LINC00847 SNORD96A MIR4638
          nsv462591 Chr.5:181195615 - 181263936 on Build GRCh38 Loss TRIM52-AS1 TRIM52 SNORD95 CTC-338M12.4 RACK1 TRIM7 TRIM41 SNORD96A MIR4638
          dgv5862n100 Chr.5:181136493 - 181229128 on Build GRCh38 Gain OR2V2 LOC102577426 TRIM7 TRIM41 MIR4638
          nsv830567 Chr.5:181103365 - 181255322 on Build GRCh38 Loss OR2V2 OR2V1 LOC102577426 TRIM52 SNORD95 CTC-338M12.4 RACK1 TRIM7 TRIM41 SNORD96A MIR4638
          nsv517697 Chr.5:181180883 - 181263936 on Build GRCh38 Loss TRIM52-AS1 LOC102577426 TRIM52 SNORD95 CTC-338M12.4 RACK1 TRIM7 TRIM41 SNORD96A MIR4638

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          protein binding
          zinc ion binding
          ligase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline