Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01497108_cn
          See other CRIP2 CNV Assays ›
          Gene Symbol
          CRIP2
          Assay Reference Genome
          Location

          Chr.14:105479504 on build GRCh38
          Cytoband
          14q32.33
          Assay ID Hs01497108_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1397

          Gene Name:

          cysteine rich protein 2

          Gene Aliases:

          CRIP, CRP2, ESP1

          Location:

          Chr.14:105472938-105480170 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 10 - Intron 10
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CRIP2 NM_001270837.1 NP_001257766.1
          NM_001270841.1 NP_001257770.1
          NM_001312.3 NP_001303.1
          NR_073081.1
          NR_073082.1
          NR_073083.1
          NR_073084.1
          NR_073085.1
          AK057837.1
          AK091845.1
          AK223251.1 BAD96971.1
          AK300092.1
          AK311402.1
          AK315757.1
          BC000434.2 AAH00434.1
          BC001931.1 AAH01931.1
          BC034151.1 AAH34151.1
          BC128101.1
          BE271979.1
          BT019911.1 AAV38714.1
          CN367966.1
          D42123.1 BAA07703.1
          HY078632.1
          U36190.1 AAB03194.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv3996n54 Chr.14:105474226 - 105488519 on Build GRCh38 Gain CRIP1 CRIP2
          nsv817647 Chr.14:105382554 - 105480505 on Build GRCh38 Gain TEX22 CRIP2 LOC100507437 PACS2 MTA1
          dgv3998n54 Chr.14:105474495 - 105770768 on Build GRCh38 Loss MIR8071-2 IGHA2 CRIP1 ELK2AP IGHG1 IGHG4 IGHG3 IGHG2 TMEM121 LOC105370697 IGH CRIP2 MIR8071-1 LOC107987211 IGHA1 C14orf80
          nsv1160204 Chr.14:105291411 - 105483783 on Build GRCh38 Duplication TEX22 BRF1 CRIP2 LOC100507437 PACS2 MTA1
          nsv566229 Chr.14:105474172 - 105483783 on Build GRCh38 Loss CRIP2
          nsv566206 Chr.14:105439736 - 105543235 on Build GRCh38 Loss TMEM121 LOC105370697 CRIP1 CRIP2 MTA1 C14orf80
          nsv832890 Chr.14:105304955 - 105483089 on Build GRCh38 Loss TEX22 BRF1 CRIP2 LOC100507437 PACS2 MTA1
          nsv1440 Chr.14:105473170 - 105479862 on Build GRCh38 Insertion CRIP2
          nsv566210 Chr.14:105465248 - 105488368 on Build GRCh38 Gain CRIP1 CRIP2 MTA1
          nsv9177 Chr.14:105476516 - 105485998 on Build GRCh38 Gain CRIP2
          nsv827088 Chr.14:105454469 - 105498549 on Build GRCh38 Gain CRIP1 CRIP2 MTA1 C14orf80
          nsv952282 Chr.14:105363764 - 105510163 on Build GRCh38 Deletion TEX22 CRIP1 CRIP2 LOC100507437 PACS2 MTA1 C14orf80
          dgv3999n54 Chr.14:105474867 - 105487998 on Build GRCh38 Loss CRIP1 CRIP2
          esv2760014 Chr.14:105047009 - 105483089 on Build GRCh38 Loss TEX22 NUDT14 MIR6765 BRF1 CRIP2 JAG2 BTBD6 LOC102723354 LOC100507437 GPR132 PACS2 MTA1
          nsv470679 Chr.14:105124547 - 105605978 on Build GRCh38 Loss IGHA2 BRF1 CRIP1 JAG2 LOC100507437 MTA1 TMEM121 TEX22 NUDT14 MIR6765 LOC105370697 IGH CRIP2 BTBD6 PACS2 C14orf80
          dgv3987n54 Chr.14:105461599 - 105499970 on Build GRCh38 Loss CRIP1 CRIP2 MTA1 C14orf80
          nsv1054190 Chr.14:105246882 - 105701138 on Build GRCh38 Gain MIR8071-2 IGHA2 BRF1 CRIP1 ELK2AP IGHG4 LOC100507437 MTA1 IGHG2 TMEM121 TEX22 LOC105370697 IGH CRIP2 BTBD6 MIR8071-1 PACS2 C14orf80
          esv3635645 Chr.14:105473682 - 105484540 on Build GRCh38 Gain CRIP2
          nsv517055 Chr.14:105136282 - 106779068 on Build GRCh38 Gain+Loss MIR8071-2 IGHA2 IGHM BRF1 JAG2 ELK2AP IGHG1 IGHV1-69 IGHG4 KIAA0125 MIR4537 IGHG2 TMEM121 TEX22 NUDT14 MIR6765 LOC105370697 IGH BTBD6 MIR8071-1 LOC107987211 IGHV3-41 IGHV3-23 IGHV3-69-1 LINC00226 IGHV3-71 C14orf80 IGHV3-30 IGHD CRIP1 MIR4507 LOC100507437 LINC00221 MIR4539 IGHG3 MTA1 IGHV4-31 ADAM6 IGHV4-55 CRIP2 IGHA1 PACS2 IGHV3-48 MIR4538
          dgv3991n54 Chr.14:105472502 - 105480505 on Build GRCh38 Loss CRIP2

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          hemopoiesis
          zinc ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline