Search
Search

Entrez Gene ID: | 5598 |
Gene Name: | mitogen-activated protein kinase 7 |
Gene Aliases: |
BMK1, ERK4, ERK5, PRKM7 |
Location: |
Chr.17:19377750-19383544 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK7 | NM_002749.4 | 2 | 290 | NP_002740.2 |
| NM_139032.3 | NP_620601.1 | |||
| NM_139033.3 | 2 | 509 | NP_620602.2 | |
| NM_139034.3 | 2 | 202 | NP_620603.2 | |
| XM_006721557.4 | 2 | 290 | XP_006721620.1 | |
| XM_006721558.4 | 2 | 509 | XP_006721621.1 | |
| XM_006721559.4 | 2 | 202 | XP_006721622.1 | |
| XM_011523957.4 | XP_011522259.1 | |||
| XM_047436401.1 | 1 | 283 | XP_047292357.1 | |
| XM_047436402.1 | 2 | 386 | XP_047292358.1 | |
| XM_047436403.1 | 2 | 298 | XP_047292359.1 | |
| XM_047436404.1 | 1 | 555 | XP_047292360.1 | |
| XM_047436405.1 | 2 | 298 | XP_047292361.1 | |
| XM_047436406.1 | 2 | 386 | XP_047292362.1 | |
| XM_047436407.1 | 2 | 290 | XP_047292363.1 | |
| AB209611.1 | 1 | 524 | BAD92848.1 | |
| AK295376.1 | 2 | 232 | BAG58335.1 | |
| AY534741.1 | 1 | 165 | AAS38577.1 | |
| BC007404.2 | AAH07404.1 | |||
| BC007992.2 | AAH07992.1 | |||
| BC009963.2 | 2 | 259 | AAH09963.1 | |
| BC030134.1 | 2 | 174 | AAH30134.1 | |
| BP871947.1 | 2 | 233 | ||
| DA452383.1 | 2 | 327 | ||
| DB133227.1 | ||||
| U25278.1 | 2 | 235 | AAA81381.1 | |
| U29725.1 | 2 | 373 | AAA82931.1 | |
| U29726.1 | 2 | 206 | AAA82932.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4235211 | Chr.17:19248750 - 19538890 on Build GRCh38 | Duplication |
|
| nsv4247486 | Chr.17:19371687 - 19382787 on Build GRCh38 | Duplication |
|
| esv3582561 | Chr.17:19240831 - 19585268 on Build GRCh38 | Loss |
|
| nsv1061317 | Chr.17:19047542 - 20322438 on Build GRCh38 | Gain |
|
| nsv953845 | Chr.17:19377788 - 19387787 on Build GRCh38 | Deletion |
|
Set Membership: |
|