Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01565980_cn
          See other GRID1 CNV Assays ›
          Gene Symbol
          GRID1
          Assay Reference Genome
          Location

          Chr.10:85600262 on build GRCh38
          Cytoband
          Assay ID Hs01565980_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          2894

          Gene Name:

          glutamate ionotropic receptor delta type subunit 1

          Gene Aliases:

          GluD1, GluD1-b

          Location:

          Chr.10:85599552-86366795 on Build GRCh38

          Assay Gene Location:

          Within Exon 18
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          GRID1 NM_017551.3 16 5444 NP_060021.1
          XM_047425122.1 9 3914 XP_047281078.1
          XM_047425123.1 9 4123 XP_047281079.1
          AB033046.1 13 4389 BAA86534.1
          BK014583.1 17 5218

          Target Gene Details

          Entrez Gene ID:

          100507470

          Gene Name:

          GRID1 antisense RNA 1

          Gene Aliases:

          -

          Location:

          Chr.10:85577731-85607215 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          GRID1-AS1 NR_038986.1
          AK097655.1
          OA985799.1
          OA985800.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3891878 Chr.10:85560815 - 85628886 on Build GRCh38 Loss GRID1 GRID1-AS1
          nsv1052427 Chr.10:85234813 - 86270793 on Build GRCh38 Gain GRID1-AS2 LINC01520 GRID1 LINC02647 MIR346 LOC101929662 GRID1-AS1

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transport
          phospholipase C-activating G-protein coupled receptor signaling pathway
          negative regulation of synaptic plasticity
          ion transmembrane transport
          social behavior
          ionotropic glutamate receptor signaling pathway
          synaptic transmission, glutamatergic
          regulation of synaptic plasticity
          modulation of synaptic transmission
          regulation of postsynaptic membrane potential
          regulation of postsynaptic membrane neurotransmitter receptor levels
          regulation of postsynapse organization
          synaptic signaling via neuropeptide
          AMPA glutamate receptor activity
          ion channel activity
          protein binding
          ligand-gated ion channel activity
          GABA receptor activity
          signaling receptor activity
          identical protein binding
          G protein-coupled receptor activity involved in regulation of postsynaptic membrane potential
          transmitter-gated ion channel activity involved in regulation of postsynaptic membrane potential

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline