Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01651511_cn
          See other AMFR CNV Assays ›
          Gene Symbol
          AMFR
          Assay Reference Genome
          Location

          Chr.16:56385966 on build GRCh38
          Cytoband
          Assay ID Hs01651511_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          267

          Gene Name:

          autocrine motility factor receptor

          Gene Aliases:

          GP78, RNF45, SPG89

          Location:

          Chr.16:56361452-56425545 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 13 - Exon 13
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          AMFR NM_001144.6 NP_001135.3
          NM_001323511.2 NP_001310440.1
          NM_001323512.2 NP_001310441.1
          XM_005255890.5 XP_005255947.1
          AF124145.1 AAD56722.1
          AK023874.1
          BC017043.1 AAH17043.1
          BC056869.1 AAH56869.1
          BC069197.1 AAH69197.1
          BI766313.1
          BQ889409.1
          CD642692.1
          L35233.1 AAA79362.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv833242 Chr.16:56259354 - 56446840 on Build GRCh38 Loss NUDT21 GNAO1 AMFR
          nsv833243 Chr.16:56375371 - 56549461 on Build GRCh38 Gain OGFOD1 NUDT21 BBS2 AMFR

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          ubiquitin-dependent protein catabolic process
          signal transduction
          I-kappaB kinase/NF-kappaB signaling
          learning or memory
          Wnt signaling pathway
          protein ubiquitination
          endoplasmic reticulum unfolded protein response
          ERAD pathway
          NIK/NF-kappaB signaling
          protein K27-linked ubiquitination
          protein autoubiquitination
          protein K48-linked ubiquitination
          negative regulation of canonical Wnt signaling pathway
          endoplasmic reticulum mannose trimming
          regulation of SREBP signaling pathway
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          transferase activity
          protein binding, bridging
          ubiquitin-ubiquitin ligase activity
          signaling receptor activity
          identical protein binding
          ubiquitin binding
          metal ion binding
          chaperone binding
          ubiquitin protein ligase activity
          BAT3 complex binding
          ubiquitin-specific protease binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline