Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01778767_cn
          See other PPIP5K1 CNV Assays ›
          Gene Symbol
          PPIP5K1
          Assay Reference Genome
          Location

          Chr.15:43534380 on build GRCh38
          Cytoband
          Assay ID Hs01778767_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          9677

          Gene Name:

          diphosphoinositol pentakisphosphate kinase 1

          Gene Aliases:

          HISPPD2A, IP6K, IPS1, VIP1, hsVIP1

          Location:

          Chr.15:43533475-43590253 on Build GRCh38

          Assay Gene Location:

          Within Exon 36
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PPIP5K1 NM_001130858.4 31 4755 NP_001124330.1
          NM_001130859.3 29 4720 NP_001124331.1
          NM_001190214.2 30 4719 NP_001177143.1
          NM_001354382.3 31 4743 NP_001341311.1
          NM_001354383.2 31 4877 NP_001341312.1
          NM_001354384.2 30 4751 NP_001341313.1
          NM_001354385.2 30 4783 NP_001341314.1
          NM_001354386.2 31 4768 NP_001341315.1
          NM_001354387.2 31 4764 NP_001341316.1
          NM_001354388.2 30 4759 NP_001341317.1
          NM_001354389.2 31 4744 NP_001341318.1
          NM_001354390.2 29 4688 NP_001341319.1
          NM_001354391.2 29 4631 NP_001341320.1
          NM_001354392.2 30 4782 NP_001341321.1
          NM_001354393.2 30 4701 NP_001341322.1
          NM_001354394.2 30 4641 NP_001341323.1
          NM_001354395.2 28 4562 NP_001341324.1
          NM_001354396.2 28 4505 NP_001341325.1
          NM_001354397.2 28 4594 NP_001341326.1
          NM_001354398.2 29 4672 NP_001341327.1
          NM_001354399.2 29 4729 NP_001341328.1
          NM_001354400.2 29 4741 NP_001341329.1
          NM_001354401.2 31 6303 NP_001341330.1
          NM_001354402.2 30 6175 NP_001341331.1
          NM_001393969.1 31 4830 NP_001380898.1
          NM_001393970.1 31 4919 NP_001380899.1
          NM_001393971.1 31 4806 NP_001380900.1
          NM_001394395.1 32 4926 NP_001381324.1
          NM_014659.6 29 4643 NP_055474.3
          XM_005254804.1 30 4725 XP_005254861.1
          XM_006720786.3 29 4599 XP_006720849.1
          XM_011522249.1 32 4938 XP_011520551.1
          XM_011522250.2 32 4918 XP_011520552.1
          XM_011522252.2 31 4933 XP_011520554.1
          XM_011522254.2 31 4844 XP_011520556.1
          XM_011522258.2 25 4087 XP_011520560.1
          XM_017022736.2 32 4975 XP_016878225.1
          XM_017022737.2 31 4913 XP_016878226.1
          XM_017022747.2 29 4667 XP_016878236.1
          XM_017022753.2 29 4696 XP_016878242.1
          XM_017022754.2 29 4688 XP_016878243.1
          XM_047433376.1 32 5027 XP_047289332.1
          XM_047433377.1 31 4901 XP_047289333.1
          XM_047433378.1 32 5249 XP_047289334.1
          XM_047433379.1 32 4914 XP_047289335.1
          XM_047433380.1 30 4781 XP_047289336.1
          XM_047433381.1 30 4694 XP_047289337.1
          XM_047433382.1 31 4845 XP_047289338.1
          XM_047433383.1 31 4853 XP_047289339.1
          XM_047433384.1 30 4670 XP_047289340.1
          XM_047433385.1 30 4814 XP_047289341.1
          XM_047433386.1 30 4790 XP_047289342.1
          XM_047433387.1 29 4607 XP_047289343.1
          XM_047433388.1 29 4579 XP_047289344.1
          XM_047433389.1 29 4656 XP_047289345.1
          XM_047433390.1 29 4575 XP_047289346.1
          AB002375.1 29 4641 BAA20831.3
          AF502586.1 30 4681 AAP30842.1
          AF502587.1 29 4647 AAP30843.1
          AF502588.1 29 4615 AAP30844.1
          AF502589.1 28 4005 AAP30845.1
          AF543190.1 28 3955 AAN40768.1
          BC057395.2 30 4722 AAH57395.1
          BX538022.1 29 4671 CAD97968.1
          BX647814.1 30 4677 CAI46011.1
          BX647870.1 8 1896

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6496110 Chr.15:43175921 - 43650339 on Build GRCh38 Duplication PPIP5K1 PPIP5K1P1-CATSPER2 ZSCAN29 CKMT1B TGM5 STRC MAPDA CCNDBP1 LCMT2 TMEM62 TUBGCP4 TGM7 TP53BP1 CATSPER2 MAP1A EPB42
          nsv832988 Chr.15:43512443 - 43684664 on Build GRCh38 Loss PPIP5K1 PPIP5K1P1-CATSPER2 CATSPER2 MAP1A CKMT1B STRC
          nsv518707 Chr.15:43509826 - 43534359 on Build GRCh38 Loss PPIP5K1 TP53BP1 MAP1A
          nsv437799 Chr.15:43491802 - 43697320 on Build GRCh38 Loss PPIP5K1 TP53BP1 PPIP5K1P1-CATSPER2 CKMT1A CATSPER2 MAP1A CKMT1B STRC
          nsv428304 Chr.15:43530960 - 43716382 on Build GRCh38 Loss PPIP5K1 STRCP1 PPIP5K1P1-CATSPER2 CKMT1A CATSPER2 MAP1A CKMT1B STRC

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          inositol metabolic process
          inositol phosphate biosynthetic process
          inositol phosphate metabolic process
          nucleotide binding
          inositol-1,3,4,5,6-pentakisphosphate kinase activity
          inositol hexakisphosphate kinase activity
          inositol heptakisphosphate kinase activity
          inositol hexakisphosphate 5-kinase activity
          ATP binding
          kinase activity
          transferase activity
          diphosphoinositol-pentakisphosphate kinase activity
          inositol hexakisphosphate 1-kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline