Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01922973_cn
          See other PFKL CNV Assays ›
          Gene Symbol
          PFKL
          Assay Reference Genome
          Location

          Chr.21:44316341 on build GRCh38
          Cytoband
          Assay ID Hs01922973_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5211

          Gene Name:

          phosphofructokinase, liver type

          Gene Aliases:

          ATP-PFK, PFK-B, PFK-L

          Location:

          Chr.21:44300053-44327373 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 14 - Intron 14
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PFKL NM_001002021.3 NP_001002021.2
          NM_002626.6 NP_002617.3
          XM_005261135.4 XP_005261192.1
          XM_005261137.5 XP_005261194.1
          XM_006724011.2 XP_006724074.1
          XM_011529603.3 XP_011527905.1
          XM_017028369.2 XP_016883858.1
          XM_024452085.2 XP_024307853.1
          XM_047440822.1 XP_047296778.1
          XM_047440823.1 XP_047296779.1
          XM_047440824.1 XP_047296780.1
          XM_047440825.1 XP_047296781.1
          AB209127.1 BAD92364.1
          AK091488.1
          AK303605.1 BAG64620.1
          AK308437.1
          BC006422.1 AAH06422.1
          BC007536.1 AAH07536.1
          BC008964.2 AAH08964.1
          BC009919.2 AAH09919.1
          BC018295.1 AAH18295.2
          BU177678.1
          BX537446.1 CAD97688.1
          X15573.1 CAA33597.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4276300 Chr.21:44310747 - 44317909 on Build GRCh38 Deletion PFKL
          nsv953646 Chr.21:44279118 - 44523117 on Build GRCh38 Deletion LRRC3 AIRE TRPM2 TSPEAR-AS1 PFKL LINC02575 TRPM2-AS CFAP410 TSPEAR TSPEAR-AS2 LRRC3-DT
          nsv1058359 Chr.21:44304172 - 44547662 on Build GRCh38 Gain LRRC3 TRPM2 TSPEAR-AS1 PFKL LINC02575 TRPM2-AS KRTAP10-1 CFAP410 TSPEAR TSPEAR-AS2 LRRC3-DT
          esv2723638 Chr.21:44249174 - 44593709 on Build GRCh38 Deletion LRRC3 TSPEAR-AS1 PFKL KRTAP10-4 TSPEAR DNMT3L KRTAP10-2 AIRE DNMT3L-AS1 TRPM2 LINC02575 TRPM2-AS KRTAP10-3 KRTAP10-5 KRTAP10-1 CFAP410 KRTAP10-6 TSPEAR-AS2 LRRC3-DT
          nsv509800 Chr.21:44284048 - 44471571 on Build GRCh38 Insertion LRRC3 AIRE TRPM2 PFKL TRPM2-AS CFAP410 LRRC3-DT
          nsv6041254 Chr.21:44287180 - 44349861 on Build GRCh38 Deletion AIRE PFKL CFAP410
          nsv587779 Chr.21:44300477 - 44330463 on Build GRCh38 Loss PFKL CFAP410
          nsv587740 Chr.21:44209682 - 44429981 on Build GRCh38 Loss AIRE DNMT3L-AS1 TRPM2 PFKL TRPM2-AS ICOSLG CFAP410 DNMT3L
          nsv6553846 Chr.21:44306061 - 44353372 on Build GRCh38 Deletion TRPM2 PFKL CFAP410
          esv33817 Chr.21:43749531 - 44647625 on Build GRCh38 Loss LRRC3 KRTAP10-8 AGPAT3 KRTAP10-9 TSPEAR AIRE TRPM2 KRTAP10-5 KRTAP10-10 TRAPPC10 GATD3 LRRC3-DT RRP1 KRTAP10-11 PDXK TSPEAR-AS1 PFKL PWP2 AATBC KRTAP10-4 DNMT3L KRTAP10-2 CSTB DNMT3L-AS1 KRTAP10-7 LINC02575 TRPM2-AS KRTAP10-3 ICOSLG KRTAP10-1 CFAP410 LINC01678 KRTAP10-6 TSPEAR-AS2 LOC105372832
          nsv587780 Chr.21:44302559 - 44344514 on Build GRCh38 Loss PFKL CFAP410
          nsv4283103 Chr.21:43950580 - 44658488 on Build GRCh38 Duplication LRRC3 KRTAP12-4 KRTAP10-8 AGPAT3 KRTAP10-9 TSPEAR AIRE TRPM2 KRTAP10-5 KRTAP10-10 TRAPPC10 GATD3 KRTAP12-3 LRRC3-DT KRTAP10-11 TSPEAR-AS1 PFKL PWP2 KRTAP10-4 DNMT3L KRTAP10-2 DNMT3L-AS1 KRTAP10-7 LINC02575 TRPM2-AS KRTAP10-3 ICOSLG KRTAP10-1 CFAP410 LINC01678 KRTAP10-6 TSPEAR-AS2 LOC105372832

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          fructose 6-phosphate metabolic process
          glycolytic process
          response to glucose
          fructose 1,6-bisphosphate metabolic process
          negative regulation of insulin secretion
          glycolytic process through fructose-6-phosphate
          canonical glycolysis
          nucleotide binding
          catalytic activity
          6-phosphofructokinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          kinase binding
          identical protein binding
          metal ion binding
          monosaccharide binding
          fructose binding
          fructose-6-phosphate binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline