Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01935322_cn
          See other FBXO24 CNV Assays ›
          Gene Symbol
          FBXO24
          Assay Reference Genome
          Location

          Chr.7:100600831 on build GRCh38
          Cytoband
          7q22.1
          Assay ID Hs01935322_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          26261

          Gene Name:

          F-box protein 24

          Gene Aliases:

          FBX24

          Location:

          Chr.7:100582490-100601117 on Build GRCh38

          Assay Gene Location:

          Within Exon 14
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          FBXO24 NM_001163499.1 10 1707 NP_001156971.1
          NM_012172.4 10 1857 NP_036304.2
          NM_033506.2 10 1968 NP_277041.1
          XM_005250259.3 9 1971 XP_005250316.1
          XM_011516022.1 10 1700 XP_011514324.1
          XM_011516023.2 9 1959 XP_011514325.1
          XM_017011961.1 11 3443 XP_016867450.1
          AK292240.1 10 1975
          AK301864.1 10 1801
          AK302252.1 10 1707
          AL136811.1 10 1913 CAB66745.1
          BC067293.1 10 1908 AAH67293.1

          Target Gene Details

          Entrez Gene ID:

          100129845

          Gene Name:

          PCOLCE antisense RNA 1

          Gene Aliases:

          -

          Location:

          Chr.7:100589401-100604038 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 3 - Exon 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PCOLCE-AS1 NR_038910.1
          AK097705.1
          AK298216.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2659884 Chr.7:100600622 - 100605969 on Build GRCh38 Deletion FBXO24 PCOLCE PCOLCE-AS1
          esv2734888 Chr.7:99975623 - 101290285 on Build GRCh38 Deletion SRRT PCOLCE-AS1 ACTL6B ZASP MIR106B MIR6840 FBXO24 PCOLCE TFR2 GNB2 MIR4653 POP7 AZGP1 MCM7 GIGYF1 AP1S1 GATS TAF6 UFSP1 MBLAC1 EPHB4 PILRA NYAP1 STAG3 LAMTOR4 TRIM56 AGFG2 SERPINE1 SAP25 AZGP1P1 COPS6 ZSCAN21 SLC12A9 GAL3ST4 CNPY4 AP4M1 PVRIG2P NAT16 MUC17 FIS1 LOC102724094 STAG3L5P MIR4658 MIR6875 PPP1R35 ZAN TRIP6 PMS2P1 GPC2 ZKSCAN1 STAG3L5P-PVRIG2P-PILRB VGF LRCH4 MEPCE TSC22D4 SPDYE3 C7orf43 PVRIG PILRB ZNF3 ZNHIT1 PLOD3 MOSPD3 LOC105375429 CLDN15 MIR93 ACHE EPO ZCWPW1 MUC12 MIR25 MUC3A MOGAT3 C7orf61
          nsv831072 Chr.7:100564880 - 100765212 on Build GRCh38 Loss PCOLCE-AS1 ACTL6B ZASP MOSPD3 LOC105375429 AGFG2 SAP25 FBXO24 ZAN PCOLCE TFR2 GNB2 EPO POP7 GIGYF1 LRCH4
          nsv951386 Chr.7:100566478 - 100682977 on Build GRCh38 Deletion PCOLCE-AS1 ACTL6B ZASP MOSPD3 LOC105375429 AGFG2 SAP25 FBXO24 PCOLCE TFR2 GNB2 GIGYF1 LRCH4
          nsv5868 Chr.7:100572375 - 100617023 on Build GRCh38 Deletion SAP25 FBXO24 PCOLCE PCOLCE-AS1 ZASP MOSPD3 LRCH4

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          ubiquitin-protein transferase activity
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline