Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01967947_cn
          See other SELENBP1 CNV Assays ›
          Gene Symbol
          SELENBP1
          Assay Reference Genome
          Location

          Chr.1:151365237 on build GRCh38
          Cytoband
          Assay ID Hs01967947_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          8991

          Gene Name:

          selenium binding protein 1

          Gene Aliases:

          EHMTO, HEL-S-134P, LPSB, MTO, SBP56, SP56, hSBP

          Location:

          Chr.1:151364304-151372705 on Build GRCh38

          Assay Gene Location:

          Within Exon 10
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SELENBP1 NM_001258288.2 9 967 NP_001245217.1
          NM_001258289.2 10 1362 NP_001245218.1
          NM_003944.4 10 1153 NP_003935.2
          XM_047433576.1 9 1183 XP_047289532.1
          AK023875.1 6 2273 BAB14709.1
          AK296661.1 10 1360 BAG59258.1
          AK298355.1 10 1047 BAG60599.1
          AK303815.1 9 962 BAG64765.1
          AK315643.1 10 1152 BAG38010.1
          BC009084.1 10 1136 AAH09084.1
          BC032997.1 8 1567 AAH32997.1
          BQ045424.1 3 587
          CR456852.1 9 1089 CAG33133.1
          EU794660.1 10 1136 ACJ13714.1
          U29091.1 9 1093 AAB02395.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv522840 Chr.1:150961247 - 151375270 on Build GRCh38 Gain SCNM1 PRUNE1 GABPB2 LYSMD1 PI4KB MLLT11 MINDY1 PSMD4 ZNF687-AS1 TMOD4 SETDB1 C1orf56 RFX5 BNIPL TNFAIP8L2 TNFAIP8L2-SCNM1 PIP5K1A SELENBP1 CERS2 CDC42SE1 VPS72 ANXA9 RFX5-AS1 SEMA6C ZNF687
          dgv53e214 Chr.1:151359619 - 151441151 on Build GRCh38 Gain POGZ PSMB4 SELENBP1
          nsv8380 Chr.1:151357411 - 151437870 on Build GRCh38 Gain POGZ PSMB4 SELENBP1
          nsv831581 Chr.1:151316675 - 151506129 on Build GRCh38 Loss RFX5-AS1 POGZ RFX5 PI4KB PSMB4 SELENBP1
          nsv441714 Chr.1:151364223 - 151425402 on Build GRCh38 Gain POGZ PSMB4 SELENBP1
          esv3587526 Chr.1:151360740 - 151441151 on Build GRCh38 Loss POGZ PSMB4 SELENBP1
          esv2758969 Chr.1:151356318 - 151439159 on Build GRCh38 Gain POGZ PSMB4 SELENBP1
          esv3587525 Chr.1:151360256 - 151412906 on Build GRCh38 Gain POGZ PSMB4 SELENBP1
          nsv824575 Chr.1:151359057 - 151435620 on Build GRCh38 Gain POGZ PSMB4 SELENBP1
          nsv4067263 Chr.1:151359592 - 151435987 on Build GRCh38 Duplication POGZ PSMB4 SELENBP1
          esv34461 Chr.1:151268066 - 151439159 on Build GRCh38 Gain ZNF687-AS1 RFX5-AS1 POGZ RFX5 PI4KB ZNF687 PSMB4 SELENBP1
          nsv1161560 Chr.1:151359541 - 151436260 on Build GRCh38 Duplication POGZ PSMB4 SELENBP1
          dgv76n145 Chr.1:151358895 - 151432320 on Build GRCh38 Duplication POGZ PSMB4 SELENBP1
          nsv5981509 Chr.1:151359872 - 151436237 on Build GRCh38 Duplication POGZ PSMB4 SELENBP1
          esv3894200 Chr.1:151360691 - 151435217 on Build GRCh38 Gain POGZ PSMB4 SELENBP1
          nsv438026 Chr.1:151346257 - 151372227 on Build GRCh38 Loss RFX5-AS1 RFX5 SELENBP1
          nsv4062004 Chr.1:151356470 - 151369416 on Build GRCh38 Duplication SELENBP1
          nsv6138677 Chr.1:151359589 - 151436025 on Build GRCh38 Duplication POGZ PSMB4 SELENBP1

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs72710112] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          protein transport
          protein binding
          selenium binding
          oxidoreductase activity
          methanethiol oxidase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline