Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01982377_cn
          See other SYNGAP1 CNV Assays ›
          Gene Symbol
          SYNGAP1
          Assay Reference Genome
          Location

          Chr.6:33440809 on build GRCh38
          Cytoband
          6p21.32
          Assay ID Hs01982377_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          8831

          Gene Name:

          synaptic Ras GTPase activating protein 1

          Gene Aliases:

          MRD5, RASA1, RASA5, SYNGAP

          Location:

          Chr.6:33420070-33453689 on Build GRCh38

          Assay Gene Location:

          Within Exon 11
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SYNGAP1 NM_006772.2 11 1952 NP_006763.2
          AK124666.1
          AK307888.1 11 1767
          AL713634.1 4 608 CAD28452.1
          CN412954.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3608630 Chr.6:33439052 - 34076935 on Build GRCh38 Gain BAK1 ITPR3 LOC105375026 LINC00336 GGNBP1 ZBTB9 MIR3934 MLN IP6K3 MIR1275 MIR7159 SYNGAP1 LOC101929188 LEMD2 GRM4 LINC01016 UQCC2
          nsv830637 Chr.6:33361588 - 33542927 on Build GRCh38 Loss KIFC1 MIR5004 PHF1 CUTA SYNGAP1 ZBTB9
          nsv1019212 Chr.6:33430254 - 33446153 on Build GRCh38 Gain MIR5004 SYNGAP1
          nsv830635 Chr.6:33270228 - 33446010 on Build GRCh38 Loss RPS18 KIFC1 ZBTB22 MIR5004 PHF1 RGL2 DAXX TAPBP VPS52 CUTA SYNGAP1 PFDN6 MIR6834 WDR46 B3GALT4 MIR6873
          nsv1073969 Chr.6:31456422 - 33480523 on Build GRCh38 Deletion NEU1 SAPCD1-AS1 HCG24 TAP1 DDX39B C6orf25 HLA-DRA GPANK1 DDAH2 LY6G5C C6orf48 ZBTB9 NFKBIL1 VPS52 TAP2 SNORD84 MIR6721 HLA-DMB LY6G6E NOTCH4 ZBTB22 LOC102725068 ZBTB12 PPT2-EGFL8 HLA-DQB1-AS1 LTA AIF1 SLC39A7 BRD2 RXRB PBX2 SNORA38 MIR219A1 SNORD117 DXO LOC100294145 SNORD48 HSPA1B MIR1236 LST1 AGER HLA-DPB2 HLA-DPB1 PPP1R2P1 TNXA MSH5 CYP21A2 HLA-DQB2 TNF LOC101929163 C6orf10 HLA-DQA1 MIR6832 HLA-DQB1 HSPA1A SKIV2L LY6G6C CYP21A1P DAXX APOM LY6G6F EHMT2 LOC100507547 C4B PSMB9 HLA-DPA1 HLA-DRB1 PSMB8 RPS18 LOC100287329 PSMB8-AS1 HCP5 RNF5 PRRT1 ATP6V1G2-DDX39B HLA-DOB ATF6B C6orf47 DDX39B-AS1 SYNGAP1 ABHD16A VARS LSM2 AGPAT1 MICB MIR5004 C4A HCG23 MIR6833 SLC44A4 LTB TAPBP CFB CSNK2B COL11A2 SNORD52 SAPCD1 HCG25 HLA-DMA MSH5-SAPCD1 CLIC1 WDR46 HLA-DRB6 HLA-DRB5 HLA-DQA2 PRRC2A PPT2 STK19 KIFC1 NCR3 HLA-DOA PHF1 EGFL8 C2 MCCD1 NELFE HSPA1L B3GALT4 VWA7 RGL2 BTNL2 HSD17B8 MIR3135B C2-AS1 ATP6V1G2 GPSM3 FKBPL LY6G6D CUTA LY6G5B PFDN6 HCG26 RING1 MIR6834 BAG6 TNXB MIR4646 MIR6873
          nsv1112900 Chr.6:31455323 - 33480423 on Build GRCh38 Deletion NEU1 SAPCD1-AS1 HCG24 TAP1 DDX39B C6orf25 HLA-DRA GPANK1 DDAH2 LY6G5C C6orf48 ZBTB9 NFKBIL1 VPS52 TAP2 SNORD84 MIR6721 HLA-DMB LY6G6E NOTCH4 ZBTB22 LOC102725068 ZBTB12 PPT2-EGFL8 HLA-DQB1-AS1 LTA AIF1 SLC39A7 BRD2 RXRB PBX2 SNORA38 MIR219A1 SNORD117 DXO LOC100294145 SNORD48 HSPA1B MIR1236 LST1 AGER HLA-DPB2 HLA-DPB1 PPP1R2P1 TNXA MSH5 CYP21A2 HLA-DQB2 TNF LOC101929163 C6orf10 HLA-DQA1 MIR6832 HLA-DQB1 HSPA1A SKIV2L LY6G6C CYP21A1P DAXX APOM LY6G6F EHMT2 LOC100507547 C4B PSMB9 HLA-DPA1 HLA-DRB1 PSMB8 RPS18 LOC100287329 PSMB8-AS1 HCP5 RNF5 PRRT1 ATP6V1G2-DDX39B HLA-DOB ATF6B C6orf47 DDX39B-AS1 SYNGAP1 ABHD16A VARS LSM2 AGPAT1 MICB MIR5004 C4A HCG23 MIR6833 SLC44A4 LTB TAPBP CFB CSNK2B COL11A2 SNORD52 SAPCD1 HCG25 HLA-DMA MSH5-SAPCD1 CLIC1 WDR46 HLA-DRB6 HLA-DRB5 HLA-DQA2 PRRC2A PPT2 STK19 KIFC1 NCR3 HLA-DOA PHF1 EGFL8 C2 MCCD1 NELFE HSPA1L B3GALT4 VWA7 RGL2 BTNL2 HSD17B8 MIR3135B C2-AS1 ATP6V1G2 GPSM3 FKBPL LY6G6D CUTA LY6G5B PFDN6 HCG26 RING1 MIR6834 BAG6 TNXB MIR4646 MIR6873
          nsv437503 Chr.6:33440765 - 33453800 on Build GRCh38 Loss SYNGAP1
          nsv1119076 Chr.6:32595823 - 33480623 on Build GRCh38 Deletion RPS18 KIFC1 HCG24 PSMB8-AS1 HLA-DOA PHF1 TAP1 HLA-DPB2 HLA-DPB1 PPP1R2P1 ZBTB9 VPS52 HLA-DOB TAP2 SYNGAP1 HLA-DQB2 B3GALT4 HLA-DMB HLA-DQA1 ZBTB22 MIR5004 HLA-DQB1 RGL2 HLA-DQB1-AS1 HSD17B8 DAXX MIR3135B TAPBP SLC39A7 BRD2 COL11A2 RXRB CUTA MIR219A1 PSMB9 HCG25 HLA-DMA PFDN6 RING1 MIR6834 HLA-DPA1 WDR46 LOC100294145 PSMB8 MIR6873 HLA-DQA2
          nsv602832 Chr.6:33414464 - 33552444 on Build GRCh38 Loss MIR5004 PHF1 CUTA SYNGAP1 ZBTB9

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          Ras protein signal transduction
          pattern specification process
          visual learning
          dendrite development
          receptor clustering
          regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          positive regulation of GTPase activity
          negative regulation of Ras protein signal transduction
          regulation of synaptic plasticity
          regulation of long-term neuronal synaptic plasticity
          negative regulation of axonogenesis
          GTPase activator activity
          SH3 domain binding
          protein kinase binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline