Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02024119_cn
          See other PPIP5K1 CNV Assays ›
          Gene Symbol
          PPIP5K1
          Assay Reference Genome
          Location

          Chr.15:43534148 on build GRCh38
          Cytoband
          Assay ID Hs02024119_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          9677

          Gene Name:

          diphosphoinositol pentakisphosphate kinase 1

          Gene Aliases:

          HISPPD2A, IP6K, IPS1, VIP1, hsVIP1

          Location:

          Chr.15:43533475-43590253 on Build GRCh38

          Assay Gene Location:

          Within Exon 36
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PPIP5K1 NM_001130858.4 31 4987 NP_001124330.1
          NM_001130859.3 29 4952 NP_001124331.1
          NM_001190214.2 30 4951 NP_001177143.1
          NM_001354382.3 31 4975 NP_001341311.1
          NM_001354383.2 31 5109 NP_001341312.1
          NM_001354384.2 30 4983 NP_001341313.1
          NM_001354385.2 30 5015 NP_001341314.1
          NM_001354386.2 31 5000 NP_001341315.1
          NM_001354387.2 31 4996 NP_001341316.1
          NM_001354388.2 30 4991 NP_001341317.1
          NM_001354389.2 31 4976 NP_001341318.1
          NM_001354390.2 29 4920 NP_001341319.1
          NM_001354391.2 29 4863 NP_001341320.1
          NM_001354392.2 30 5014 NP_001341321.1
          NM_001354393.2 30 4933 NP_001341322.1
          NM_001354394.2 30 4873 NP_001341323.1
          NM_001354395.2 28 4794 NP_001341324.1
          NM_001354396.2 28 4737 NP_001341325.1
          NM_001354397.2 28 4826 NP_001341326.1
          NM_001354398.2 29 4904 NP_001341327.1
          NM_001354399.2 29 4961 NP_001341328.1
          NM_001354400.2 29 4973 NP_001341329.1
          NM_001354401.2 31 6535 NP_001341330.1
          NM_001354402.2 30 6407 NP_001341331.1
          NM_001393969.1 31 5062 NP_001380898.1
          NM_001393970.1 31 5151 NP_001380899.1
          NM_001393971.1 31 5038 NP_001380900.1
          NM_001394395.1 32 5158 NP_001381324.1
          NM_014659.6 29 4875 NP_055474.3
          XM_005254804.1 30 4957 XP_005254861.1
          XM_006720786.3 29 4831 XP_006720849.1
          XM_011522249.1 32 5170 XP_011520551.1
          XM_011522250.2 32 5150 XP_011520552.1
          XM_011522252.2 31 5165 XP_011520554.1
          XM_011522254.2 31 5076 XP_011520556.1
          XM_011522258.2 25 4319 XP_011520560.1
          XM_017022736.2 32 5207 XP_016878225.1
          XM_017022737.2 31 5145 XP_016878226.1
          XM_017022747.2 29 4899 XP_016878236.1
          XM_017022753.2 29 4928 XP_016878242.1
          XM_017022754.2 29 4920 XP_016878243.1
          XM_047433376.1 32 5259 XP_047289332.1
          XM_047433377.1 31 5133 XP_047289333.1
          XM_047433378.1 32 5481 XP_047289334.1
          XM_047433379.1 32 5146 XP_047289335.1
          XM_047433380.1 30 5013 XP_047289336.1
          XM_047433381.1 30 4926 XP_047289337.1
          XM_047433382.1 31 5077 XP_047289338.1
          XM_047433383.1 31 5085 XP_047289339.1
          XM_047433384.1 30 4902 XP_047289340.1
          XM_047433385.1 30 5046 XP_047289341.1
          XM_047433386.1 30 5022 XP_047289342.1
          XM_047433387.1 29 4839 XP_047289343.1
          XM_047433388.1 29 4811 XP_047289344.1
          XM_047433389.1 29 4888 XP_047289345.1
          XM_047433390.1 29 4807 XP_047289346.1
          AB002375.1 29 4873 BAA20831.3
          AF502586.1 30 4913 AAP30842.1
          AF502588.1 29 4847 AAP30844.1
          AF502589.1 28 4237 AAP30845.1
          AF543190.1 28 4187 AAN40768.1
          BC057395.2 30 4954 AAH57395.1
          BX538022.1 29 4903 CAD97968.1
          BX647814.1 30 4909 CAI46011.1
          BX647870.1 8 2128

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6496110 Chr.15:43175921 - 43650339 on Build GRCh38 Duplication MAP1A TP53BP1 CKMT1B TMEM62 CATSPER2 PPIP5K1 PPIP5K1P1-CATSPER2 EPB42 STRC TGM5 ZSCAN29 MAPDA TUBGCP4 CCNDBP1 TGM7 LCMT2
          nsv832988 Chr.15:43512443 - 43684664 on Build GRCh38 Loss MAP1A CKMT1B CATSPER2 PPIP5K1 PPIP5K1P1-CATSPER2 STRC
          nsv518707 Chr.15:43509826 - 43534359 on Build GRCh38 Loss MAP1A TP53BP1 PPIP5K1
          nsv437799 Chr.15:43491802 - 43697320 on Build GRCh38 Loss MAP1A TP53BP1 CKMT1B CATSPER2 PPIP5K1 PPIP5K1P1-CATSPER2 STRC CKMT1A
          nsv428304 Chr.15:43530960 - 43716382 on Build GRCh38 Loss MAP1A CKMT1B CATSPER2 STRCP1 PPIP5K1 PPIP5K1P1-CATSPER2 STRC CKMT1A

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          inositol metabolic process
          inositol phosphate biosynthetic process
          inositol phosphate metabolic process
          nucleotide binding
          inositol-1,3,4,5,6-pentakisphosphate kinase activity
          inositol hexakisphosphate kinase activity
          inositol heptakisphosphate kinase activity
          inositol hexakisphosphate 5-kinase activity
          ATP binding
          kinase activity
          transferase activity
          diphosphoinositol-pentakisphosphate kinase activity
          inositol hexakisphosphate 1-kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline