Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02124641_cn
          See other RAD51C CNV Assays ›
          Gene Symbol
          RAD51C
          Assay Reference Genome
          Location

          Chr.17:58732515 on build GRCh38
          Cytoband
          17q22
          Assay ID Hs02124641_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5889

          Gene Name:

          RAD51 paralog C

          Gene Aliases:

          BROVCA3, FANCO, R51H3, RAD51L2

          Location:

          Chr.17:58692538-58734342 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 10 - Intron 10
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          RAD51C NM_058216.2 NP_478123.1
          NR_103872.1
          XM_006722001.3 XP_006722064.1
          XM_006722002.3 XP_006722065.1
          XM_006722004.3 XP_006722067.1
          XM_006722005.3 XP_006722068.1
          XM_011525092.2 XP_011523394.1
          XM_011525093.2 XP_011523395.1
          XM_011525094.2 XP_011523396.1
          XM_017024914.1 XP_016880403.1
          XM_017024915.1 XP_016880404.1
          XM_017024916.1 XP_016880405.1
          XM_017024917.1 XP_016880406.1
          XM_017024918.1 XP_016880407.1
          XM_017024919.1 XP_016880408.1
          AF029669.1 AAC39604.1
          AK303363.1
          BC107753.1 AAI07754.1
          CB243543.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv833498 Chr.17:58652063 - 58815031 on Build GRCh38 Loss RAD51C TEX14 PPM1E

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          DNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance via recombination
          double-strand break repair via homologous recombination
          DNA recombinase assembly
          DNA synthesis involved in DNA repair
          strand displacement
          DNA repair
          DNA recombination
          mitotic recombination
          sister chromatid cohesion
          female meiosis sister chromatid cohesion
          reciprocal meiotic recombination
          male meiosis I
          spermatogenesis
          blood coagulation
          response to ionizing radiation
          positive regulation of G2/M transition of mitotic cell cycle
          strand invasion
          chromosome organization involved in meiotic cell cycle
          recombinase activity
          four-way junction DNA binding
          DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          protein binding
          ATP binding
          DNA-dependent ATPase activity
          crossover junction endodeoxyribonuclease activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline