Search
Search

Entrez Gene ID: | 56924 |
Gene Name: | p21 (RAC1) activated kinase 6 |
Gene Aliases: |
PAK5 |
Location: |
Chr.15:40239091-40277487 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAK6 | NM_001276717.1 | NP_001263646.1 | ||
| NM_001276718.1 | 10 | 2821 | NP_001263647.1 | |
| NM_020168.5 | 11 | 2543 | NP_064553.1 | |
| AF276893.1 | 10 | 2242 | AAF82800.1 | |
| AJ236915.1 | 11 | 2483 | CAC18720.1 | |
| AK131522.1 | 10 | 2821 | ||
| BC035596.1 | AAH35596.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471240 | Chr.15:40235954 - 40303426 on Build GRCh38 | Loss |
|
| nsv510660 | Chr.15:40260705 - 40429684 on Build GRCh38 | Deletion |
|
| nsv569227 | Chr.15:40273504 - 40314283 on Build GRCh38 | Loss |
|
| esv34145 | Chr.15:40257416 - 40597626 on Build GRCh38 | Loss |
|
| nsv517541 | Chr.15:40265067 - 40303426 on Build GRCh38 | Loss |
|
| nsv952611 | Chr.15:40237800 - 40284699 on Build GRCh38 | Deletion |
|